Merck
Molecular Logic and Evolution of Bitter Taste in Drosophila.
Current biology : CB (2019-12-17)
Hany K M Dweck, John R Carlson
ABSTRACT
Taste systems detect a vast diversity of toxins, which are perceived as bitter. When a species adapts to a new environment, its taste system must adapt to detect new death threats. We deleted each of six commonly expressed bitter gustatory receptors (Grs) from Drosophila melanogaster. Systematic analysis revealed that requirements for these Grs differed for the same tastant in different neurons and for different tastants in the same neuron. Responses to some tastants in some neurons required four Grs, including Gr39a. Deletions also produced increased or novel responses, supporting a model of Gr-Gr inhibitory interactions. Coexpression of four Grs conferred several bitter responses to a sugar neuron. We then examined bitter coding in three other Drosophila species. We found major evolutionary shifts. One shift depended on the concerted activity of seven Grs. This work shows how the complex logic of bitter coding provides the capacity to detect innumerable hazards and the flexibility to adapt to new ones.
Product Description
Supelco
Myricetin, analytical standard
Sigma-Aldrich
Azadirachtin, ~95%
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
(−)-Lobeline hydrochloride, 98%
Supelco
Rotenone, PESTANAL®, analytical standard
Sigma-Aldrich
Quinine, suitable for fluorescence, anhydrous, ≥98.0% (dried material, NT)
Sigma-Aldrich
Escin, ≥95%, powder
Berberine chloride, European Pharmacopoeia (EP) Reference Standard
Supelco
DEET, PESTANAL®, analytical standard
Sigma-Aldrich
Sucrose octaacetate, ≥97%, FG
Sigma-Aldrich
Coumarin, ≥99% (HPLC)
Sigma-Aldrich
(±)-Gossypol from cotton seeds, ≥95% (HPLC)
Sigma-Aldrich
Theobromine, ≥98.0%
Sigma-Aldrich
Denatonium benzoate, ≥98%
Sigma-Aldrich
Theophylline, anhydrous, ≥99%, powder
Sigma-Aldrich
Aristolochic acid I, powder
Sigma-Aldrich
Cucurbitacin I hydrate, ≥95% (HPLC), solid
Sigma-Aldrich
Saponin, for molecular biology, used as non-ionic surfactant