Skip to Content
Merck
All Photos(1)

Key Documents

C1778

Sigma-Aldrich

Caffeine

Sigma Reference Standard, vial of 250 mg

Synonym(s):

1,3,7-Trimethylxanthine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10N4O2
CAS Number:
Molecular Weight:
194.19
Beilstein:
17705
EC Number:
MDL number:
UNSPSC Code:
12352210
eCl@ss:
34050301
PubChem Substance ID:
NACRES:
NA.21
grade:
Sigma Reference Standard
Pricing and availability is not currently available.

grade

Sigma Reference Standard

Quality Level

form

crystalline
powder

packaging

vial of 250 mg

mp

234-236.5 °C (lit.)

solubility

H2O: soluble 18.7 g/L at 16 °C

SMILES string

CN1C(=O)N(C)c2ncn(C)c2C1=O

InChI

1S/C8H10N4O2/c1-10-4-9-6-5(10)7(13)12(3)8(14)11(6)2/h4H,1-3H3

InChI key

RYYVLZVUVIJVGH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Supplied in amber screw-cap vials

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

CONSORT extension for reporting N-of-1 trials (CENT) 2015: Explanation and elaboration.
Larissa Shamseer et al.
BMJ (Clinical research ed.), 350, h1793-h1793 (2015-05-16)
France Denoeud et al.
Science (New York, N.Y.), 345(6201), 1181-1184 (2014-09-06)
Coffee is a valuable beverage crop due to its characteristic flavor, aroma, and the stimulating effects of caffeine. We generated a high-quality draft genome of the species Coffea canephora, which displays a conserved chromosomal gene order among asterid angiosperms. Although
Nicolas Bousette et al.
Journal of cellular physiology, 229(3), 374-383 (2013-09-17)
Calnexin (CNX) is an endoplasmic reticulum (ER) quality control chaperone that has been implicated in ER stress. ER stress is a prominent pathological feature of various pathologic conditions, including cardiovascular diseases. However, the role of CNX and ER stress has
Louise A Horrigan et al.
Pharmacology & therapeutics, 111(3), 877-892 (2006-03-17)
Caffeine is a member of the methylxanthine family of drugs, and is the most widely consumed behaviourally active substance in the western world. This article is focused on the impact of caffeine on immune function. In this regard, a number
Jonathan M Hodgson et al.
The American journal of clinical nutrition, 97(5), 943-950 (2013-04-05)
Measures of blood pressure variation have been associated with cardiovascular disease and related outcomes. The regular consumption of black tea can lower blood pressure, but its effects on blood pressure variation have yet to be investigated. We aimed to assess

Articles

Validated HPLC method (based on Ph. Eur. Monograph 2668) for quantifying catechins and caffeine in decaffeinated green tea using a Chromolith® High Resolution RP-18e column with advantages in terms of backpressure and matrix robustness.

Validated HPLC method (based on Ph. Eur. Monograph 2668) for quantifying catechins and caffeine in decaffeinated green tea using a Chromolith® High Resolution RP-18e column with advantages in terms of backpressure and matrix robustness.

Validated HPLC method (based on Ph. Eur. Monograph 2668) for quantifying catechins and caffeine in decaffeinated green tea using a Chromolith® High Resolution RP-18e column with advantages in terms of backpressure and matrix robustness.

Validated HPLC method (based on Ph. Eur. Monograph 2668) for quantifying catechins and caffeine in decaffeinated green tea using a Chromolith® High Resolution RP-18e column with advantages in terms of backpressure and matrix robustness.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service