Skip to Content
Merck
All Photos(1)

Documents

Y0001149

Berberine chloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Berberine chloride form, Natural Yellow 18

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H18ClNO4
CAS Number:
Molecular Weight:
371.81
Colour Index Number:
75160
Beilstein:
3836585
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

berberine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COC1=C(C=[N+](CCC2=C3C=C4C(OCO4)=C2)C3=C5)C5=CC=C1OC.[Cl-]

InChI

1S/C20H18NO4.ClH/c1-22-17-4-3-12-7-16-14-9-19-18(24-11-25-19)8-13(14)5-6-21(16)10-15(12)20(17)23-2;/h3-4,7-10H,5-6,11H2,1-2H3;1H/q+1;/p-1

InChI key

VKJGBAJNNALVAV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Fluorescent stain for heparin in mast cells

Biochem/physiol Actions

An alkaloid with weak antibiotic properties. Substrate for MDR efflux pumps. Antimicrobial activities of berberine is potentiated by the MDR inhibitor 5´-methoxyhydnocarpin (5´-MHC). Berberine upregulates the expression of Pgp in hepatoma cells. Treatment with berberine potentially results in the reduced accumulation of chemotherapeutic drugs.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of berberine on amelioration of hyperglycemia and oxidative stress in high glucose and high fat diet-induced diabetic hamsters in vivo.
Liu C, Wang Z, Song Y, et al.
BioMed Research International, 2015, 313808-313808 (2015)
Mohamed A Dkhil et al.
Parasitology research, 114(4), 1581-1593 (2015-02-11)
Plant-based natural products are promising sources for identifying novel agents with potential anti-Eimeria activity. This study explores possible effects of berberine on Eimeria papillata infections in the jejunum of male Swiss albino mice. Berberine chloride, when daily administered to mice
Victor P Bagla et al.
BMC complementary and alternative medicine, 14, 383-383 (2014-10-09)
Different parts of Podocarpus henkelii have been used in many cultures around the world to treat ailments such as cholera, stomach diseases, rheumatism, cancer, canine distemper in dogs and gall sickness in cattle. The aim of this study was to
Mousa O Germoush et al.
Journal of cancer research and clinical oncology, 140(7), 1103-1109 (2014-04-20)
The present study was designed to investigate the possible protective effects of berberine against cyclophosphamide (CP)-induced hepatotoxicity in rats. The experimental rats were treated with berberine orally at a dose of 50 mg/kg for 11 consecutive days after the administration
Hany K M Dweck et al.
Current biology : CB, 30(1), 17-30 (2019-12-17)
Taste systems detect a vast diversity of toxins, which are perceived as bitter. When a species adapts to a new environment, its taste system must adapt to detect new death threats. We deleted each of six commonly expressed bitter gustatory receptors (Grs)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service