Skip to Content
Merck
All Photos(1)

Key Documents

22620

Sigma-Aldrich

Quinine

suitable for fluorescence, anhydrous, ≥98.0% (dried material, NT)

Synonym(s):

6′-Methoxycinchonidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24N2O2
CAS Number:
Molecular Weight:
324.42
Beilstein:
91867
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.32
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (dried material, NT)

form

powder

optical activity

[α]20/D −126±5°, c = 1% in chloroform

impurities

≤5% dihydroquinine (HPLC)

loss

≤1% loss on drying, 110 °C

mp

173-175 °C (lit.)

solubility

H2O: soluble

fluorescence

λex 347 nm; λem 448 nm in 0.5 M sulfuric acid

suitability

suitable for fluorescence

SMILES string

COc1ccc2nccc([C@@H](O)[C@@H]3C[C@@H]4CCN3C[C@@H]4C=C)c2c1

InChI

1S/C20H24N2O2/c1-3-13-12-22-9-7-14(13)10-19(22)20(23)16-6-8-21-18-5-4-15(24-2)11-17(16)18/h3-6,8,11,13-14,19-20,23H,1,7,9-10,12H2,2H3/t13-,14-,19-,20+/m0/s1

InChI key

LOUPRKONTZGTKE-WZBLMQSHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Quinine, also known as 6′-Methoxycinchonidine is a fluorescent reagent. The quantum yield of Quinine is 23% higher at 390 mµ excitation wavelength than at 313 mµ. The fluorescence polarization in the emission band of quinine in a rigid medium arises from two singlet states simultaneously. The emission spectra of quinine or 6-methoxyquinoline shifts towards the red zone when excited at 390 mµ.[1]

Application

Quinine was used in the following processes:

  • To study its in vitro antimalarial activity in combination with omeprazole.[2]
  • To analyze its effect on viscosity and friction of saliva.[3]
  • As a test agent to study its impact on the accumulation of the fluorescent P-glycoprotein (Pgp) substrates in P-glycoprotein overexpressing breast cancer cells.[4]
  • To study its influence on the pyramidal cell intrinsic properties, extracellular potassium transients, and epileptiform activity in vitro.[5]
  • As a reference compound to identify alkaloids by phytochemical screening of Deianira erubescens, Strychnos pseudoquina and Remijia ferruginea plants.[6]

Biochem/physiol Actions

Potassium channel blocker

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The role of friction in perceived oral texture.
Rene A. de Wijk , Jon F. Prinz.
Food Quality and Preference, 16, 121-129 (2005)
Jeffrey G Sarver et al.
Journal of biomolecular screening, 7(1), 29-34 (2002-03-19)
A microplate screening method has been developed to evaluate the effects of test agents on the accumulation of the fluorescent P-glycoprotein (Pgp) substrates Hoechst 33342, rhodamine 123, and rhodamine 6G in multidrug-resistant (MDR) breast cancer cells that overexpress Pgp. All
V F Andrade-Neto et al.
Journal of ethnopharmacology, 87(2-3), 253-256 (2003-07-16)
For centuries, malaria was treated with the bark of Cinchona calisaya and Cinchona succirubra plants named "quinas" in Brazil, from which the quinine molecule was isolated. Other plant species known also as "quinas" are used to treat fever and malaria
Some characteristics of the fluorescence of quinine.
R F Chen
Analytical biochemistry, 19(2), 374-387 (1967-05-01)
T Skinner-Adams et al.
Antimicrobial agents and chemotherapy, 43(5), 1304-1306 (1999-05-01)
Previous studies have shown that the proton pump inhibitor omeprazole has antimalarial activity in vitro. The interactions of omeprazole with commonly used antimalarial drugs were assessed in vitro. Omeprazole and quinine combinations were synergistic; however, chloroquine and omeprazole combinations were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service