Skip to Content
Merck
All Photos(1)

Key Documents

1672745

USP

Tranexamic acid

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

trans-4-(Aminomethyl)cyclohexanecarboxylic acid, AMCA, AMCHA, HAKU, TAMCHA, Tranexamic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCH2C6H10CO2H
CAS Number:
Molecular Weight:
157.21
Beilstein:
2207452
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

tranexamic acid

manufacturer/tradename

USP

mp

>300 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

NC[C@H]1CC[C@@H](CC1)C(O)=O

InChI

1S/C8H15NO2/c9-5-6-1-3-7(4-2-6)8(10)11/h6-7H,1-5,9H2,(H,10,11)/t6-,7-

InChI key

GYDJEQRTZSCIOI-LJGSYFOKSA-N

Gene Information

human ... PLG(5340)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Tranexamic acid USP Reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Tranexamic Acid Tablets
  • Tranexamic Acid Injection

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chih-Hsiang Chang et al.
Clinical orthopaedics and related research, 472(5), 1552-1557 (2014-01-05)
Systemic tranexamic acid can decrease blood loss and rates of transfusion in patients undergoing total hip arthroplasty (THA). However, the efficacy of topical tranexamic acid in THA has only recently been characterized in a small number of studies. The purpose
Baohui Yang et al.
PloS one, 8(2), e55436-e55436 (2013-02-21)
Tranexamic acid (TXA) is well-established as a versatile oral, intramuscular, and intravenous (IV) antifibrinolytic agent. However, the efficacy of IV TXA in reducing perioperative blood transfusion in spinal surgery is poorly documented. We conducted a meta-analysis of randomized controlled trials
Zhi-Jun Li et al.
European spine journal : official publication of the European Spine Society, the European Spinal Deformity Society, and the European Section of the Cervical Spine Research Society, 22(9), 1950-1957 (2013-05-10)
The present meta-analysis aimed at assessing the effectiveness and safety of tranexamic acid (TXA) in reducing blood loss and transfusion in spinal surgery. Systematic searches of all studies published through March 2012 were identified from PubMed, EMBase, Cochrane library, Science
K Ker et al.
The British journal of surgery, 100(10), 1271-1279 (2013-07-11)
Tranexamic acid (TXA) reduces blood transfusion in surgery but the extent of the reduction in blood loss and how it relates to the dose of TXA is unclear. A systematic review of randomized trials was performed. Data were extracted on
Anshul Dahuja et al.
The Journal of arthroplasty, 29(4), 733-735 (2013-11-05)
Total knee arthroplasty(TKA) is associated with extensive postoperative blood loss. Despite various studies proving the efficacy of Tranexamic Acid (TEA) with single or multiple boluses of different sizes with or without subsequent infusions, no consensus has been reached on the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service