Skip to Content
Merck
All Photos(4)

Key Documents

857653

Sigma-Aldrich

trans-4-(Aminomethyl)cyclohexanecarboxylic acid

97%

Synonym(s):

AMCA, AMCHA, HAKU, TAMCHA, Tranexamic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCH2C6H10CO2H
CAS Number:
Molecular Weight:
157.21
Beilstein:
2207452
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

>300 °C (lit.)

SMILES string

NC[C@H]1CC[C@@H](CC1)C(O)=O

InChI

1S/C8H15NO2/c9-5-6-1-3-7(4-2-6)8(10)11/h6-7H,1-5,9H2,(H,10,11)/t6-,7-

InChI key

GYDJEQRTZSCIOI-LJGSYFOKSA-N

Gene Information

human ... PLG(5340)

Looking for similar products? Visit Product Comparison Guide

Application

Used as a lysine analogue to characterize binding sites in plasminogen.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Thomas Forro et al.
Cerebral cortex (New York, N.Y. : 1991), 25(5), 1228-1240 (2013-11-28)
Travelling theta oscillations and sharp wave-associated ripples (SWRs) provide temporal structures to neural activity in the CA1 hippocampus. The contribution of rhythm-generating GABAergic interneurons to network timing across the septotemporal CA1 axis remains unknown. We recorded the spike-timing of identified
Case scenario: management of trauma-induced coagulopathy in a severe blunt trauma patient.
Jean-Stephane David et al.
Anesthesiology, 119(1), 191-200 (2013-03-22)
J M Marshall et al.
Biochemistry, 33(12), 3599-3606 (1994-03-29)
The conformations of Glu-plasminogen and defined proteolytic fragments, in the presence and absence of 6-aminohexanoic acid (6-AHA), trans-4-(aminomethyl)cyclohexanecarboxylic acid (t-AMCHA), and benzamidine, were studied using three methods: size-exclusion high-performance liquid chromatography (SE-HPLC), small-angle X-ray scattering (SAXS), and dynamic laser light
G J Hoover et al.
Biochemistry, 32(41), 10936-10943 (1993-10-19)
A series of strategically designed recombinant (r) mutants of the kringle 1 region of human plasminogen ([K1HPg]) have been constructed and the resulting gene products employed to reveal the identities of the residues that contribute to stabilization of the binding
Kristen A Matteson et al.
Obstetrics and gynecology, 121(3), 632-643 (2013-05-03)
To compare the effectiveness of nonsurgical abnormal uterine bleeding treatments for bleeding control, quality of life (QOL), pain, sexual health, patient satisfaction, additional treatments needed, and adverse events. MEDLINE, Cochrane databases, and Clinicaltrials.gov were searched from inception to May 2012.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service