Skip to Content
Merck
All Photos(1)

Key Documents

Y0002098

Sorafenib

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4-[4-[[[[4-Chloro-3-(trifluoromethyl)phenyl]amino]carbonyl]amino]phenoxy]-N-methyl-2-pyridinecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H16ClF3N4O3
CAS Number:
Molecular Weight:
464.82
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

sorafenib

manufacturer/tradename

EDQM

application(s)

pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

FC(F)(F)c1c(ccc(c1)NC(=O)Nc2ccc(cc2)Oc3cc(ncc3)C(=O)NC)Cl

InChI

1S/C21H16ClF3N4O3/c1-26-19(30)18-11-15(8-9-27-18)32-14-5-2-12(3-6-14)28-20(31)29-13-4-7-17(22)16(10-13)21(23,24)25/h2-11H,1H3,(H,26,30)(H2,28,29,31)

InChI key

MLDQJTXFUGDVEO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sorafenib EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - Repr. 1B - STOT RE 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yiliang Wang et al.
Journal of nanobiotechnology, 22(1), 473-473 (2024-08-13)
Hepatocellular carcinoma (HCC) is a highly malignant tumor known for its hypoxic environment, which contributes to resistance against the anticancer drug Sorafenib (SF). Addressing SF resistance in HCC requires innovative strategies to improve tumor oxygenation and effectively deliver therapeutics. In
Zhong Wan et al.
Aging, 12(18), 18192-18208 (2020-10-07)
We investigated MAPK14-dependent resistance to sorafenib in hepatocellular carcinoma (HCC). Bioinformatics analysis and dual luciferase reporter assays in HCC cell lines showed that miR-216a-3p directly binds to the 3'UTR of MAPK14 mRNA and downregulates MAPK14 protein expression. Consequently, miR-216a-3p expression
Ye Kuang et al.
Cancer medicine, 12(6), 7222-7233 (2022-11-25)
Hepatocellular carcinoma (HCC) is a primary liver cancer with high mortality. The long-term use of sorafenib, a targeted drug for hepatocellular carcinoma, will lead to drug resistance, and patients are prone to cancer metastasis, the molecular mechanism of which is
Baoxin Zhao et al.
Virologica Sinica, 38(5), 680-689 (2023-06-19)
Chronic hepatitis B virus (HBV) infection is one of the leading causes of hepatocellular carcinoma (HCC). The HBV genome is prone to mutate and several variants are closely related to the malignant transformation of liver disease. G1896A mutation (G to
Su-Hong Chen et al.
Experimental and therapeutic medicine, 23(3), 232-232 (2022-03-01)
Sorafenib is currently used to treat hepatocellular carcinoma (HCC). However, the development of chemoresistance to sorafenib is a major limitation for sorafenib-based therapy in patients with HCC. In the present study, the effect of the combination therapy of sorafenib and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service