Skip to Content
Merck
All Photos(1)

Key Documents

SML3017

Sigma-Aldrich

Lenvatinib mesylate

≥98% (HPLC)

Synonym(s):

4-[3-Chloro-4-(N’-cyclopropylureido)phenoxy]-7-methoxyquinoline-6-carboxamide methanesulfonate, 4-[3-Chloro-4-[(cyclopropylaminocarbonyl)amino]phenoxy]-7-methoxy-6-quinolinecarboxamide mesylate, 4-[3-chloro-4-[[(cyclopropylamino)carbonyl]amino]phenoxy]-7-methoxy-6-Quinolinecarboxamide methanesulfonate (1:1), E 7080 mesylate, E-7080 mesylate, E7080 mesylate, ER-203492-00 mesylate, Lenvatinib methanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H19ClN4O4 · CH4O3S
CAS Number:
Molecular Weight:
522.96
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1=CC2=C(OC3=CC=C(C(Cl)=C3)NC(NC4CC4)=O)C=CN=C2C=C1OC)N.O=S(O)(C)=O

InChI

1S/C21H19ClN4O4.CH4O3S/c1-29-19-10-17-13(9-14(19)20(23)27)18(6-7-24-17)30-12-4-5-16(15(22)8-12)26-21(28)25-11-2-3-11;1-5(2,3)4/h4-11H,2-3H2,1H3,(H2,23,27)(H2,25,26,28);1H3,(H,2,3,4)

InChI key

HWLFIUUAYLEFCT-UHFFFAOYSA-N

Biochem/physiol Actions

Lenvatinib (E7080) is an orally active inhibitor against multiple receptor tyrosine kinases, including VEGFR (Flt-1/KDR/Flt-4 IC50 = 22/4.0/5.2 nM), PDGFR1/2 (IC50 = 39/51 nM), FGFR1 and KIT (IC50 = 46 and 100 nM, respectively) that inhibits angiogenesis in vitro (VEGF/SCF-induced HUVEC tube formation IC50 = 5.1/5.2 nM). Lenvatinib oral administration results in tumor growth arrest (30 mg/kg b.i.d. p.o.) and even regression (100 mg/kg b.i.d. p.o.) in vivo among mice harboring SCF-producing human small cell lung carcinoma H146 cells.
Orally active receptor tyrosine kinase inhibitor against VEGFR (Flt-1/KDR/Flt-4), PDGFR1/2, FGFR1 and KIT with anti- angiogenesis efficacy in vitro and in vivo.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - STOT RE 2

Target Organs

Gastro-intestinal system,Kidney,Bone,Reproductive organs,Adrenal gland

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haijing Deng et al.
Liver cancer, 9(3), 338-357 (2020-07-11)
Combining anti-angiogenic therapy with immune checkpoint blockade with anti-programmed cell death-1 (PD-1) antibodies is a promising treatment for hepatocellular carcinoma (HCC). Tyrosine kinase inhibitors are well-known anti-angiogenic agents and offer potential for combination with anti-PD-1 antibodies. This study investigated the
Mariko Aoyama et al.
Oncology reports, 44(4), 1709-1716 (2020-09-19)
Anaplastic thyroid carcinoma (ATC) is a rare type of thyroid carcinoma with a poor prognosis. Thus, suitable preclinical tumor models are required for the development of new ATC therapies. In the present study, orthotopic tumor xenograft models were established using
Reina Sasaki et al.
International journal of molecular sciences, 21(9) (2020-05-14)
Multiple kinase inhibitors are available for patients with advanced hepatocellular carcinoma (HCC). It is largely unknown whether regorafenib or lenvatinib modulates innate immunity including Toll-like receptor (TLR)-signaling pathways in HCC. We performed real-time RT-PCR to investigate 84 TLR-associated gene expression
Guoqing Tang et al.
Journal of molecular neuroscience : MN, 70(9), 1437-1444 (2020-05-10)
The therapeutic effect of deferoxamine (DFO) for spinal cord injury (SCI) has been demonstrated in previous studies; however, the exact mechanism of action is still unclear. Here, we hypothesized that DFO ameliorates spinal cord compression by promoting neovascularization. Using an
Junji Matsui et al.
International journal of cancer, 122(3), 664-671 (2007-10-19)
E7080 is an orally active inhibitor of multiple receptor tyrosine kinases including VEGF, FGF and SCF receptors. In this study, we show the inhibitory activity of E7080 against SCF-induced angiogenesis in vitro and tumor growth of SCF-producing human small cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service