Skip to Content
Merck
All Photos(1)

Documents

SML2156

Sigma-Aldrich

Erlotinib hydrochloride

≥98% (HPLC)

Synonym(s):

774 hydrochloride, CP-358, N-(3-ethynylphenyl)-6,7-bis(2-methoxyethoxy)-4-quinazolinamine hydrochloride, OSI-744 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H23N3O4 · HCl
CAS Number:
Molecular Weight:
429.90
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

−20°C

InChI

1S/C22H23N3O4.ClH/c1-4-16-6-5-7-17(12-16)25-22-18-13-20(28-10-8-26-2)21(29-11-9-27-3)14-19(18)23-15-24-22;/h1,5-7,12-15H,8-11H2,2-3H3,(H,23,24,25);1H

InChI key

GTTBEUCJPZQMDZ-UHFFFAOYSA-N

General description

Erlotinib hydrochloride is an antineoplastic drug. It regulates mechanistic target of rapamycin (mTOR) inhibition, epidermal growth factor receptor down-regulation and epidermal interstitial transformation (EMT) suppression.

Application

Erlotinib hydrochloride has been used to activate mitochondrial fragmentation in a human PANC-1 pancreatic cancer cell line.

Biochem/physiol Actions

Erlotinib hydrochloride is an inhibitor of the receptor tyrosine kinase EGFR (epidermal growth factor receptor). It is used clinically for the treatment of non-small cell lung cancer (NSCLC) and in combination for metastatic pancreatic cancer.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service