Skip to Content
Merck
All Photos(1)

Key Documents

819686

TSKgel® BioAssist® S (7 µm) HPLC Columns

L x I.D. 5 cm × 4.6 mm, sulfonic acid phase

Synonym(s):

TSKgel® BioAssist® S HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501

Product Name

TSKgel® BioAssist® S HPLC Column, phase sulfonic acid, L × I.D. 5 cm × 4.6 mm, 7 μm particle size, PEEK hardware

material

PEEK hardware

product line

TSKgel®

feature

counter ion Na+

manufacturer/tradename

Tosoh 19686

parameter

≤2.6 M salt concentration
≤30% organic solvent
1.0 mL/min flow rate
25 bar max. pressure
4-60 °C temperature

technique(s)

HPLC: suitable

L × I.D.

5 cm × 4.6 mm

matrix

polymer particle platform

matrix active group

sulfonic acid phase

particle size

7 μm

pore size

1300 Å

operating pH

2-12(<= 1 month)
3-10(> 1 month)

separation technique

cation exchange

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel BioAssist S columns are packed with porous hydrophilic polymer beads which are surface modified with a strong cation exchanger. The resin′s large pores are very accessible even for high molecular weight proteins. This leads to higher chromatographic efficiency and binding capacity for purification. The columns are made of PEEK to reduce protein adsorption.

Physical form

Shipped in 20% Ethanol in 20 mM phosphate buffer (pH 6.5).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

BioAssist is a registered trademark of Tosoh Corporation
TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service