Skip to Content
Merck
All Photos(1)

Key Documents

C0150

Sigma-Aldrich

Cathepsin B from human placenta

lyophilized powder, ≥5 units/mg protein

Synonym(s):

Cathepsin B1

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

lyophilized powder

Quality Level

specific activity

≥5 units/mg protein

mol wt

24.5 kDa

composition

Protein, ~50% Lowry

solubility

H2O: soluble 1.0 mg/mL, clear

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... CTSB(1508)

General description

Cathepsin B is encoded by the gene CTSB and is located in the human chromosome at 8p23.1. Cathepsin B is a lysosomal proteolytic enzyme of cysteine protease family. It is present in all mammalian cells. It is essential for the intracellular protein turnover.

Application

Cathepsin B from human placenta has been used in the in vitro digestion of modified myelin oligodendrocyte glycoprotein (MOG). It has also been used in determining the effect of cysteine protease inhibitors.
Cathepsin B is a lysosomal cysteine proteinase which hydrolyzes proteins with a broad specificity for peptide bonds. Cathepsin B may be a useful tool in Alzheimer′s research, as it may have a role in the natural defense against the disease . Cathepsin B may be used to cleave procaspase 1 and procaspase 11, and to induce apoptosis in digitonin-permeabilized cells.

Biochem/physiol Actions

Cathepsin B has been found to cleave procaspase 1 and procaspase 11 and to induce apoptosis in digitonin-permeabilized cells. Translocation of cathepsin B from the cytoplasm to the nucleus contributes to bile salt induced apoptosis of rat hepatocytes. Levels of cathepsin B in PC12 cells significantly decrease 12 to 24 hours after apoptosis is induced.
Cathepsin B preferentially cleaves at the caboxyl side of Arg-Arg bonds in small molecule substrates. Cathepsin B has been shown to degrade β-amyloid precursor protein into harmless fragments . Cathepsin B inhibition has been shown to limit bone metastasis in breast cancer .

Unit Definition

One unit will liberate 1 nanomole of 7-amino-4-methylcoumarin from Z-Arg-Arg 7-amido-4-methylcoumarin per min at pH 6.0 at 40 °C.

Physical form

Lyophilized powder containing phosphate buffer salts

Preparation Note

Dissolves in water at 1 mg/mL concentration, yieliding a clear solution.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scavenger Receptor A Mediates the Clearance and Immunological Screening of MDA-Modified Antigen by M2-Type Macrophages
Warnecke A, et al.
Neuromolecular Medicine, 19(4), 463-479 (2017)
Cathepsins B, H, L and cysteine protease inhibitors in malignant prostate cell lines, primary cultured prostatic cells and prostatic tissue
Friedrich B, et al.
European Journal of Cancer, 35(1), 138-144 (1999)
Kamran Waidha et al.
Molecules (Basel, Switzerland), 26(16) (2021-08-28)
The specificity of inhibition by 6,6'-dihydroxythiobinupharidine (DTBN) on cysteine proteases was demonstrated in this work. There were differences in the extent of inhibition, reflecting active site structural-steric and biochemical differences. Cathepsin S (IC50 = 3.2 μM) was most sensitive to
Viktorija Herceg et al.
Molecules (Basel, Switzerland), 25(18) (2020-09-24)
Cyclopeptidic chemotherapeutic prodrugs (cPCPs) are macromolecular protease-sensitive doxorubicin (DOX) prodrugs synthesized from a cyclodecapeptidic scaffold, termed Regioselectively Addressable Functionalized Template (RAFT). In order to increase the chemotherapeutic potential of DOX and limit its toxicity, we used a Cathepsin B (Cat
Sarah Mueller-Steiner et al.
Neuron, 51(6), 703-714 (2006-09-20)
Alzheimer's disease (AD) may result from the accumulation of amyloid-beta (Abeta) peptides in the brain. The cysteine protease cathepsin B (CatB) is associated with amyloid plaques in AD brains and has been suspected to increase Abeta production. Here, we demonstrate

Articles

Cathepsin B is a lysosomal cysteine proteinase with broad specificity. This protocol uses Nα–CBZ–Arg–Arg–7–amido–4–methylcoumarin as the substrate for fluorometric detection of Cathepsin B activity.

Cathepsin B is a lysosomal cysteine proteinase with broad specificity. This protocol uses Nα–CBZ–Arg–Arg–7–amido–4–methylcoumarin as the substrate for fluorometric detection of Cathepsin B activity.

Cathepsin B is a lysosomal cysteine proteinase with broad specificity. This protocol uses Nα–CBZ–Arg–Arg–7–amido–4–methylcoumarin as the substrate for fluorometric detection of Cathepsin B activity.

Cathepsin B is a lysosomal cysteine proteinase with broad specificity. This protocol uses Nα–CBZ–Arg–Arg–7–amido–4–methylcoumarin as the substrate for fluorometric detection of Cathepsin B activity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service