Skip to Content
Merck
All Photos(3)

Key Documents

561223

Sigma-Aldrich

Poly(4-styrenesulfonic acid) solution

Mw ~75,000, 18 wt. % in H2O

Synonym(s):

PSS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C8H8O3S)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

mol wt

Mw ~75,000

concentration

18 wt. % in H2O

refractive index

n20/D 1.3718

pH

1.55 (25 °C)

viscosity

20 cP(25 °C, Brookfield) (20 rpm)

solubility

H2O: soluble
ethanol: soluble
lower glycols: soluble
methanol: soluble

density

1.11 g/mL at 25 °C

SMILES string

[Ca+2].[S](=O)(=O)([O-])c1c(cccc1)C=C

InChI

1S/C8H8O3S.Ca/c1-2-7-5-3-4-6-8(7)12(9,10)11;/h2-6H,1H2,(H,9,10,11);/q;+2/p-1

InChI key

IJBYVKWTVVYTPL-UHFFFAOYSA-M

Application

Polyelectrolyte. Electroconductive and antistatic resin for electrographic and electrophotographic substrates.

Features and Benefits

Excellent film-forming and coating capability. Good conductive properties over a broad range of relative humidity.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karen M Gallegos et al.
Frontiers in microbiology, 9, 3269-3269 (2019-02-01)
Chlamydia is the most frequently reported sexually transmitted bacteria causing 2.9 million infections annually in the United States. Diagnosis, treatment, and sequelae of chlamydial disease cost billions of dollars each year in the United States alone. Considering that a heparin
Naïg Aurélia Lüdmilla Chenais et al.
Journal of neural engineering (2020-11-25)
Retinal stimulation in blind patients evokes the sensation of discrete points of light called phosphenes, which allows them to perform visually guided tasks, such as orientation, navigation, object recognition, object manipulation and reading. However, the clinical benefit of artificial vision
M Bretaudeau Deguigne et al.
Clinical toxicology (Philadelphia, Pa.), 51(4), 243-248 (2013-04-10)
The objective of this study was to evaluate the effects of early digestive tract decontamination on the severity of acute-on-chronic lithium poisoning (acute poisoning in patients under long-term therapy). This was an observational and retrospective study of acute-on-chronic lithium overdoses
Swee Pin Yeap et al.
Langmuir : the ACS journal of surfaces and colloids, 28(42), 14878-14891 (2012-10-03)
A detailed study on the conflicting role that colloid stability plays in magnetophoresis is presented. Magnetic iron oxide particles (MIOPs) that were sterically stabilized via surface modification with poly(sodium 4-styrene sulfonate) of different molecular weights (i.e., 70 and 1000 kDa)
Johannes Frueh et al.
Physical chemistry chemical physics : PCCP, 15(2), 483-488 (2012-11-23)
Polyelectrolyte multilayers (PEMs) deposited on flexible supports are promising candidates for many applications ranging from controlled wettability over stimuli responsive nanovalves to lithography free surface structuring. Since many potential applications involve elongation of these films, we investigated the effect of

Articles

We present an article that discusses two applications in particular; first, using these layers as polyelectrolyte membranes to control permeability.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service