57354-U
SPME Fiber Assembly
PEG, for use with autosampler, Nitinol-core (NIT), 23 ga, 60 μm (PEG)
Synonym(s):
CW/PEG Fiber Assembly
About This Item
Recommended Products
product name
SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm(PEG, for use with autosampler
material
metal alloy fiber
plain purple hub
Quality Level
needle size
23 ga
packaging
pkg of 3 × ea
greener alternative product characteristics
Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
df
60 μm (PEG)
technique(s)
solid phase microextraction (SPME): suitable
fiber L
1 cm
matrix active group
PEG coating
application(s)
food and beverages
compatibility
for use with autosampler
greener alternative category
Looking for similar products? Visit Product Comparison Guide
General description
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Chromatograms
application for SPME, suitable for GCapplication for SPME, suitable for GCapplication for SPME, suitable for GCapplication for SPME, suitable for GCShow MoreOur team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service