Skip to Content
Merck
All Photos(4)

Key Documents

57310-U

Supelco

SPME Fiber Assembly

greener alternative

PDMS/DVB, for use with manual holder, Nitinol-core (NIT), 24 ga, 65 μm (PDMS/DVB)

Synonym(s):

SPME FIBER PK3(MANUAL),W/65UM PS-DVB COATING

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115724
NACRES:
NB.21

Product Name

SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, for use with manual holder

material

fused silica fiber
plain blue hub

Quality Level

needle size

24 ga

packaging

pkg of 3 ea

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

df

65 μm (PDMS/DVB)

technique(s)

solid phase microextraction (SPME): suitable

fiber L

1 cm

matrix active group

PDMS/DVB coating

application(s)

food and beverages

compatibility

for analyte group volatiles, amines, and nitroaromatic compounds (MW 50-300)
for use with manual holder

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This is a Solid Phase Microextraction (SPME) technology product. SPME is one of the greener technologies for sample preparation as it is solvent free extraction technology and SPME fibres are re-usable and generates as little waste as possible, thus aligns with "Waste Prevention”, and ″Safer Solvents and Auxiliaries″. Click here for more information.

Application

Volatiles, Amines, and Nitroaromatic Compounds

Footnote

This SPME fiber assembly is also available with a Nitinol (NIT) core as Cat. No. 57921-U, providing enhanced fiber reproducibility, greater fiber durability, maintained selectivity for method consistency, and core inertness to ensure analyte stability. See more at SigmaAldrich.com/NITSPME .

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marisela Pontes et al.
Food chemistry, 134(4), 2509-2520 (2013-02-28)
In this study the effect of the cultivar on the volatile profile of five different banana varieties was evaluated and determined by dynamic headspace solid-phase microextraction (dHS-SPME) combined with one-dimensional gas chromatography-mass spectrometry (1D-GC-qMS). This approach allowed the definition of
C A Groom et al.
Journal of chromatography. A, 909(1), 53-60 (2001-02-24)
The cyclic nitramine explosives hexahydro-1,3,5-trinitro-1,3,5-triazine (RDX) and octahydro-1,3,5,7-tetranitro-1,3,5,7-tetrazine (HMX) were examined in field and microcosm soil samples to determine their patterns of degradation and environmental fates. A number of analytical techniques, including solid-phase microextraction with on-fiber derivatization, gas chromatography-mass spectrometry
Jing Cheng et al.
Journal of chromatography. A, 1218(18), 2476-2482 (2011-03-23)
A novel ultrasound-assisted surfactant-enhanced emulsification microextraction (UASEME) technique has been proposed by using low-density extraction solvents. In the proposed technique, Tween 80 and cyclohexane were injected into 5-mL glass test tubes with conical bottoms, containing 5.00 mL of a water
Elodie Passeport et al.
Journal of chromatography. A, 1217(33), 5317-5327 (2010-07-14)
A solid-phase microextraction (SPME)/gas chromatography (GC)-mass spectrometry (MS) multiresidue analytical method was developed for 16 pesticides presenting different physicochemical properties including diphenyl ether, triazine, ureas, acetamides, benzofuran, thiocarbamate, pyridine carboxamides, chloronitrile, piperedine, and azoles. Optimization was achieved by means of
Noureddine Yassaa et al.
Journal of chromatography. A, 1141(1), 138-144 (2006-12-19)
In order to develop a valuable method for accurate screening of biogenic emissions from undisturbed living plants or for plant-insect interactions, solid-phase microextraction (SPME) has been combined with dynamic branch enclosure cuvettes and enantioselective GC/MS. The study was conducted at

Related Content

Solid phase microextraction (SPME) is a solvent-free sample preparation technique that uses coated fibers to extract analytes of interest from a sample before analysis by GC or HPLC.

Solid phase microextraction (SPME) is a solvent-free sample preparation technique that uses coated fibers to extract analytes of interest from a sample before analysis by GC or HPLC.

Solid phase microextraction (SPME) is a solvent-free sample preparation technique that uses coated fibers to extract analytes of interest from a sample before analysis by GC or HPLC.

Solid phase microextraction (SPME) is a solvent-free sample preparation technique that uses coated fibers to extract analytes of interest from a sample before analysis by GC or HPLC.

Chromatograms

suitable for GC, application for SPMEsuitable for GC, application for SPMEsuitable for GC, application for SPMEsuitable for GC, application for SPMEShow More

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service