Skip to Content
Merck
SPME Fiber Assembly 50/30µm DVB/CAR/PDMS needle size 24 ga, StableFlex fiber, for use with manual holder

SPME Fiber Assembly

SPME Fiber Assembly 50/30µm DVB/CAR/PDMS needle size 24 ga, StableFlex fiber, for use with manual holder

Products

Brand
Sustainability
Feature
Length (cm)
Quality Segment
Application
Color
df (µm)
Package Size
Gauge
Technique
Showing 1-20 of 27
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2