Skip to Content
Merck
All Photos(2)

Documents

S1404

Sigma-Aldrich

Sophorose

≥98% (HPLC, anhydrous)

Synonym(s):

β-D-Glc-(1→2)-D-Glc, 2-O-β-D-Glucopyranosyl-D-glucopyranose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC, anhydrous)

form

powder

technique(s)

HPLC: suitable

impurities

≤8% water (Karl Fischer)

color

white

solubility

water: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

O[C@H]([C@H]1O)[C@@H](O[C@H](CO)[C@H]1O)O[C@@H](C=O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/C12H22O11/c13-1-4(16)7(17)8(18)5(2-14)22-12-11(21)10(20)9(19)6(3-15)23-12/h2,4-13,15-21H,1,3H2/t4-,5+,6-,7-,8-,9-,10+,11-,12-/m1/s1

InChI key

PZDOWFGHCNHPQD-VNNZMYODSA-N

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Arkadij P Sinitsyn et al.
Biochemistry. Biokhimiia, 86(Suppl 1), S166-S195 (2021-04-09)
The review discusses various aspects of renewable plant biomass conversion and production of the second-generation biofuels, including the types of plant biomass, its composition and reaction ability in the enzymatic hydrolysis, and various pretreatment methods for increasing the biomass reactivity.
Mari Häkkinen et al.
Microbial cell factories, 11, 134-134 (2012-10-06)
Trichoderma reesei is a soft rot Ascomycota fungus utilised for industrial production of secreted enzymes, especially lignocellulose degrading enzymes. About 30 carbohydrate active enzymes (CAZymes) of T. reesei have been biochemically characterised. Genome sequencing has revealed a large number of
D B R K Gupta Udatha et al.
BMC systems biology, 9, 77-77 (2015-11-18)
The gene expression and secretion of fungal lignocellulolytic enzymes are tightly controlled at the transcription level using independent mechanisms to respond to distinct inducers from plant biomass. An advanced systems-level understanding of transcriptional regulatory networks is required to rationally engineer
Mercedes Ramírez-Escudero et al.
The Journal of biological chemistry, 291(46), 24200-24214 (2016-09-30)
Metagenomics has opened up a vast pool of genes for putative, yet uncharacterized, enzymes. It widens our knowledge on the enzyme diversity world and discloses new families for which a clear classification is still needed, as is exemplified by glycoside
Désiré Yapi Assoi Yapi et al.
Journal of insect science (Online), 9, 4-4 (2009-07-21)
A beta-glucosidase was purified from the digestive fluid of the palm weevil Rhynchophorus palmarum L. (Coleoptera: Curculionidae) by chromatography on anion-exchange, gel filtration, and hydrophobic interaction columns. The preparation was shown to be homogeneous on polyacrylamide gels, beta-glucosidase is a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service