Skip to Content
Merck
All Photos(1)

Key Documents

SML2312

Sigma-Aldrich

Tipranavir

≥98% (HPLC)

Synonym(s):

5-trifluoromethyl-N-[3(R)-[1-[5,6-dihydro-4-hydroxy-2-oxo-6(R)-(2-phenethyl)-6(R)-n-propyl-2H-pyran-3-yl]propyl]phenyl]-2-pyridinesulfonamide, N-[3-[(1R)-1-[(6R)-5,6-dihydro-4-hydroxy-2-oxo-6-(2-phenylethyl)-6-propyl-2H-pyran-3-yl]propyl]phenyl]-5-(trifluoromethyl)-pyridinesulfonamide, PNU 140690, PNU-140690, TPV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H33F3N2O5S
CAS Number:
Molecular Weight:
602.66
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

InChI

1S/C31H33F3N2O5S/c1-3-16-30(17-15-21-9-6-5-7-10-21)19-26(37)28(29(38)41-30)25(4-2)22-11-8-12-24(18-22)36-42(39,40)27-14-13-23(20-35-27)31(32,33)34/h5-14,18,20,25,36-37H,3-4,15-17,19H2,1-2H3/t25-,30-/m1/s1

InChI key

SUJUHGSWHZTSEU-FYBSXPHGSA-N

Biochem/physiol Actions

Tipranavir is a HIV protease inhibitor that is highly potent against a variety of HIV strains and isolates in several cell culture systems. Tipranavir inhibits the replication of viruses that are resistant to other protease inhibitors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S M Poppe et al.
Antimicrobial agents and chemotherapy, 41(5), 1058-1063 (1997-05-01)
PNU-140690 is a member of a new class of nonpeptidic human immunodeficiency virus (HIV) protease inhibitors (sulfonamide-containing 5,6-dihydro-4-hydroxy-2-pyrones) discovered by structure-based design. PNU-140690 has excellent potency against a variety of HIV type 1 (HIV-1) laboratory strains and clinical isolates, including
Manabu Aoki et al.
Journal of virology, 86(24), 13384-13396 (2012-09-28)
Tipranavir (TPV), a protease inhibitor (PI) inhibiting the enzymatic activity and dimerization of HIV-1 protease, exerts potent activity against multi-PI-resistant HIV-1 isolates. When a mixture of 11 multi-PI-resistant (but TPV-sensitive) clinical isolates (HIV(11MIX)), which included HIV(B) and HIV(C), was selected
Soo-Yon Rhee et al.
Antimicrobial agents and chemotherapy, 54(10), 4253-4261 (2010-07-28)
The effects of many protease inhibitor (PI)-selected mutations on the susceptibility to individual PIs are unknown. We analyzed in vitro susceptibility test results on 2,725 HIV-1 protease isolates. More than 2,400 isolates had been tested for susceptibility to fosamprenavir, indinavir

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service