Skip to Content
Merck
All Photos(1)

Key Documents

SML0937

Sigma-Aldrich

Darunavir

≥98% (HPLC)

Synonym(s):

TMC-114, UIC-94017, [(1R,5S,6R)-2,8-dioxabicyclo[3.3.0]oct-6-yl] N-[(2S,3R)-4- [(4-aminophenyl)sulfonyl- (2-methylpropyl)amino]-3-hydroxy-1-phenyl- butan-2-yl]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H37N3O7S
CAS Number:
Molecular Weight:
547.66
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 20 mg/mL, clear

shipped in

wet ice

storage temp.

−20°C

SMILES string

[H][C@]1([C@@H](OC(N[C@@H](CC2=CC=CC=C2)[C@@H](CN(S(C3=CC=C(N)C=C3)(=O)=O)CC(C)C)O)=O)CO4)[C@]4([H])OCC1

InChI

1S/C27H37N3O7S/c1-18(2)15-30(38(33,34)21-10-8-20(28)9-11-21)16-24(31)23(14-19-6-4-3-5-7-19)29-27(32)37-25-17-36-26-22(25)12-13-35-26/h3-11,18,22-26,31H,12-17,28H2,1-2H3,(H,29,32)/t22-,23-,24+,25-,26+/m0/s1

InChI key

CJBJHOAVZSMMDJ-HEXNFIEUSA-N

Biochem/physiol Actions

Darunavir has been sanctioned by the food and drug administration (FDA) as the first treatment of drug-resistant human immunodeficiency virus (HIV).[1]
Darunavir is a HIV protease inhibitor; antiretroviral.
Darunavir is a second-generation antiviral HIV protease inhibitor with broad spectrum activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beatriz Grinsztejn et al.
The lancet. HIV, 6(9), e588-e600 (2019-08-03)
Antiretroviral therapy (ART) management is challenging for individuals in resource-limited settings presenting for third-line treatment because of complex resistance patterns, partly due to reduced access to viral load monitoring. We aimed to evaluate use of newer antiretroviral drugs and contemporary
Michael S Saag et al.
JAMA, 320(4), 379-396 (2018-07-26)
Antiretroviral therapy (ART) is the cornerstone of prevention and management of HIV infection. To evaluate new data and treatments and incorporate this information into updated recommendations for initiating therapy, monitoring individuals starting therapy, changing regimens, and preventing HIV infection for
Ninon Taylor et al.
Journal of acquired immune deficiency syndromes (1999), 75(1), e13-e20 (2016-11-01)
Hemeoxygenase-1 (HO-1) has recently been identified as a major driver of metaflammation and obesity-related insulin resistance (IR). Drug-induced IR increases cardiovascular risk within the HIV-1-infected population receiving antiretroviral therapy (ART). We therefore investigated a possible role of HO-1 in ART-induced
Joseph J Eron et al.
AIDS (London, England), 32(11), 1431-1442 (2018-04-24)
To investigate efficacy and safety of a single-tablet regimen of darunavir/cobicistat/emtricitabine/tenofovir alafenamide (D/C/F/TAF) 800/150/200/10 mg vs. darunavir/cobicistat plus emtricitabine/tenofovir disoproxyl fumarate (TDF) (control) in antiretroviral-treatment-naive, HIV-1-infected adults. Phase-3, randomized, active-controlled, double-blind, international, multicenter, noninferiority study (NCT02431247). Seven hundred and twenty-five participants
David Wu et al.
Journal of clinical medicine, 9(6) (2020-06-21)
The current COVID-19 pandemic is caused by the novel coronavirus SARS-CoV-2. The virus causes severe respiratory symptoms which manifest disproportionately in the elderly. Currently, there are over 6.5 million cases and 380,000 deaths reported. Given the current severity of the

Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service