Skip to Content
Merck
All Photos(1)

Key Documents

SML1222

Sigma-Aldrich

Lopinavir

≥98% (HPLC)

Synonym(s):

(αS)-N-[(1S,3S,4S)-4-[[(2,6-Dimethylphenoxy)acetyl]amino]-3-hydroxy-5-phenyl-1-(phenylmethyl)pentyl]tetrahydro-α-(1-methylethyl)-2-oxo-1(2H)-

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H48N4O5
CAS Number:
Molecular Weight:
628.80
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -20 to -27°, c = 0.4 in methanol

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

N4(CCCNC4=O)[C@@H](C(C)C)C(=O)N[C@H](C[C@H](O)[C@@H](NC(=O)COc3c(cccc3C)C)Cc2ccccc2)Cc1ccccc1

InChI

1S/C37H48N4O5/c1-25(2)34(41-20-12-19-38-37(41)45)36(44)39-30(21-28-15-7-5-8-16-28)23-32(42)31(22-29-17-9-6-10-18-29)40-33(43)24-46-35-26(3)13-11-14-27(35)4/h5-11,13-18,25,30-32,34,42H,12,19-24H2,1-4H3,(H,38,45)(H,39,44)(H,40,43)/t30-,31-,32-,34-/m0/s1

InChI key

KJHKTHWMRKYKJE-SUGCFTRWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Lopinavir has been used as a ZMPSTE24 and human immunodeficiency virus protease inhibitor.[1]

Biochem/physiol Actions

Lopinavir is an antiviral HIV Protease Inhibitor. Lopinavir has insufficient bioavailability alone, so it is used in therapy in combination with Ritonavir, a HIV protease inhibitor, which inhibits cytochrome P450-3A4 (CYP3A4), a liver enzyme that normally metabolizes protease inhibitors. Lopinavir also has an ability to inhibit ZMPSTE24 (zinc metallopeptidase STE24).[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Didier K Ekouevi et al.
BMC infectious diseases, 14, 461-461 (2014-08-27)
Few data are available on antiretroviral therapy (ART) response among HIV-2 infected patients. We conducted a systematic review on treatment outcomes among HIV-2 infected patients on ART, focusing on the immunological and virological responses in adults. Data were extracted from
H L Sham et al.
Antimicrobial agents and chemotherapy, 42(12), 3218-3224 (1998-12-03)
The valine at position 82 (Val 82) in the active site of the human immunodeficiency virus (HIV) protease mutates in response to therapy with the protease inhibitor ritonavir. By using the X-ray crystal structure of the complex of HIV protease
K A Sutherland et al.
The Journal of antimicrobial chemotherapy, 70(1), 243-248 (2014-09-18)
PI susceptibility results from a complex interplay between protease and Gag proteins, with Gag showing wide variation across HIV-1 subtypes. We explored the impact of pre-treatment susceptibility on the outcome of lopinavir/ritonavir monotherapy. Treatment-naive individuals who experienced lopinavir/ritonavir monotherapy failure
Translocon Declogger Ste24 Protects against IAPP Oligomer-Induced Proteotoxicity.
Kayatekin C, et al.
Cell, 173(1), 62-73 (2018)
Shutoku Matsuyama et al.
Journal of virology, 95(1) (2020-10-16)
Here, we screened steroid compounds to obtain a drug expected to block host inflammatory responses and Middle East respiratory syndrome coronavirus (MERS-CoV) replication. Ciclesonide, an inhaled corticosteroid, suppressed the replication of MERS-CoV and other coronaviruses, including severe acute respiratory syndrome

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service