Skip to Content
Merck
All Photos(1)

Key Documents

SML2051

Sigma-Aldrich

LY2109761

≥98% (HPLC)

Synonym(s):

4-[5,6-Dihydro-2-(2-pyridinyl)-4H-pyrrolo[1,2-b]pyrazol-3-yl]-7-[2-(4-morpholinyl)ethoxy]-quinoline, LY-2109761

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H27N5O2
CAS Number:
Molecular Weight:
441.52
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

[n]21nc(c(c2CCC1)c4c5c(ncc4)cc(cc5)OCCN6CCOCC6)c3ncccc3

InChI

1S/C26H27N5O2/c1-2-9-27-22(4-1)26-25(24-5-3-11-31(24)29-26)21-8-10-28-23-18-19(6-7-20(21)23)33-17-14-30-12-15-32-16-13-30/h1-2,4,6-10,18H,3,5,11-17H2

InChI key

IHLVSLOZUHKNMQ-UHFFFAOYSA-N

Application

LY2109761 has been used as an inhibitor of type I and II transforming growth factor-beta (TGFβ) receptors to reduce the phosphorylation of SMAD2 (mothers against decapentaplegic homolog) in HepG2 cells.[1]

Biochem/physiol Actions

LY2109761 is a potent and orally active TGF-β receptor (TGFβR) type I & II dual inhibitor (IC50 = 70 and 322 nM against TGFβRI/ALK5 and TGFβRII autophosphorylation, respectively, with 4 μM ATP), inhibiting Fyn/JNK3/Lck/MKK6/SAPK2α only at much higher concentrations (58-89% inhibition at 20 μM) and exhibiting little or no potency against 37 other kinases (IC50 >20 μM). LY2109761 inhibits 0.25 ng/mL TGFβ-induced NIH/3T3 proliferation in cultures (IC50 = 210 nM; 2-hr pretreatment prior to TGFβ for 24 hrs) and suppresses human MX1 breast carcinoma xenograft tumor growth in mice in vivo (by ∼80% on day 37; 75 mg/kg p.o. bid from day 7 to 20). When administered in combination with gemcitabine (25 mg/kg/day i.p.), LY2109761 (50 mg/kg p.o. bid) is shown to significantly reduce tumor burden and spontaneous abdominal metastases in a murine model of metastatic pancreatic cancer.
LY2109761 suppresses transforming growth factor (TGF)-β1-induced fibroblasts proliferation and collagen synthesis in hypertrophic scar fibroblasts. Therefore, it is used in treating hypertrophic scars.[2]
Orally active, potent and selective TGFβRI/II dual inhibitor with anti-cancer efficacy in in cultures and in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Davide Melisi et al.
Molecular cancer therapeutics, 7(4), 829-840 (2008-04-17)
Most pancreatic cancer patients present with inoperable disease or develop metastases after surgery. Conventional therapies are usually ineffective in treating metastatic disease. It is evident that novel therapies remain to be developed. Transforming growth factor beta (TGF-beta) plays a key
Hong-Yu Li et al.
Journal of medicinal chemistry, 51(7), 2302-2306 (2008-03-05)
In our continuing effort to expand the SAR of the quinoline domain of dihydropyrrolopyrazole series, we have discovered compound 15d, which demonstrated the antitumor efficacy with oral bioavailability. This effort also demonstrated that the PK/PD in vivo target inhibition paradigm
Jia Fang et al.
Scientific reports, 7, 43851-43851 (2017-03-04)
Previous studies have shown that CD61 (integrin-β3) promotes the differentiation of human umbilical cord mesenchymal stem cells (hUC-MSCs) into germ-like cells. However, the mechanism remains unclear. In this study, we showed that overexpression of CD61 in canine adipose-derived mesenchymal stem
Markus D Lacher et al.
Cancer research, 66(3), 1648-1657 (2006-02-03)
Expression of the Coxsackie and Adenovirus Receptor (CAR) is frequently reduced in carcinomas, resulting in decreased susceptibility of such tumors to infection with therapeutic adenoviruses. Because CAR participates physiologically in the formation of tight-junction protein complexes, we examined whether molecular
Xuan Liu et al.
BioMed research international, 2017, 2613198-2613198 (2017-03-17)
JPJD was an ideal alternative traditional Chinese medicine compound in the prevention and treatment of CRC, but its underlying mechanisms has not been fully elucidated. In this study, we demonstrated in vitro that TGF-β-induced EMT promoted the invasion and metastasis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service