Skip to Content
Merck
All Photos(3)

Key Documents

Y0503

Sigma-Aldrich

Y-27632 dihydrochloride

≥98% (HPLC)

Synonym(s):

Rock inhibitor y-27632, Y-27632 2HCl, (R)-(+)-trans-4-(1-Aminoethyl)-N-(4-Pyridyl)cyclohexanecarboxamide dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H21N3O · 2HCl
CAS Number:
Molecular Weight:
320.26
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +3.0 to +5.0°, c = 1.0 in methanol

color

white to beige

solubility

H2O: 14 mg/mL

storage temp.

2-8°C

SMILES string

Cl.Cl.C[C@@H](N)[C@H]1CC[C@@H](CC1)C(=O)Nc2ccncc2

InChI

1S/C14H21N3O.2ClH/c1-10(15)11-2-4-12(5-3-11)14(18)17-13-6-8-16-9-7-13;;/h6-12H,2-5,15H2,1H3,(H,16,17,18);2*1H/t10-,11-,12-;;/m1../s1

InChI key

IDDDVXIUIXWAGJ-LJDSMOQUSA-N

Gene Information

General description

Y-27632 dihydrochloride is an effective bronchodilator and improves lung resistance (RL) induced by antigen and acetylcholine. Y-27632 controls neutrophil migration and lung edema and may be a potential agent against acute lung injury.The utilization of Y-27632 dihydrochloride, in conjunction with Accutase or other enzyme solutions, during the passaging of human embryonic stem (hES) cells has been demonstrated to enhance cell survival and significantly improve cloning efficiency
Y-27632, a Rho-kinase inhibitor, counteracts noradrenergic contractions in the penile corpus cavernosum of rabbits and humans. It helps to relax vascular and non-vascular smooth muscle in various preparations and lower blood pressure in hypertensive rats. Additionally, this inhibitor induces reversible changes in cell shape and decreases in actin stress fibers, focal adhesions, and protein phosphotyrosine staining in human trabecular meshwork (TM) cells and Schlemm′s canal cells. Y-27632 inhibits the kinase activity of both ROCK-I and ROCK-II in vitro, and this inhibition is competitively reversed by ATP, indicating that these compounds bind to the catalytic site to inhibit the kinases. Ki = 140 nM for p160ROCK.

Application

Y-27632 dihydrochloride has been used:
  • as a medium supplement in pancreatic ductal adenocarcinoma organoid culture
  • in the inhibition of Ras homolog gene family (Rho) kinase in mouse embryonic stem cells
  • in the inhibition of Rho-associated protein kinase (ROCK) in human embryonic stem cells and human induced pluripotent stem cells (iPSCs)
  • Y-27632 dihydrochloride has been used as a supplement in the E8 medium to promote in vitro differentiation of human pluripotent stem cells

Biochem/physiol Actions

Y-27632 is a highly potent, cell-permeable, selective ROCK inhibitor.
Y-27632 is highly potent, cell-permeable, selective ROCK (Rho-associated coiled coil forming protein serine/threonine kinase) inhibitor. Ki = 140 nM for p160ROCK. Y-27632 also inhibits ROCK-II with equal potency. The inhibition is competitive with respect to ATP.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Legal Information

Sold under license from Mitsubishi Pharma Corporation

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y-27632, an inhibitor of Rho-kinase, antagonizes noradrenergic contractions in the rabbit and human penile corpus cavernosum
Rees RW, et al.
British Journal of Pharmacology, 133, 455-458 (2001)
Evaluation of Y-27632, a rho-kinase inhibitor, as a bronchodilator in guinea pigs
Iizuka K, et al.
European Journal of Pharmacology, 406(2), 273-279 (2000)
Reversal of diabetes with insulin-producing cells derived in vitro from human pluripotent stem cells
Rezania A, et al.
Nature Biotechnology, 32(11), 1121-1133 (2014)
Basement membrane destruction by pancreatic stellate cells leads to local invasion in pancreatic ductal adenocarcinoma
Koikawa K, et al.
Cancer Letters, 425, 65-77 (2018)
Pharmacological properties of Y-27632, a specific inhibitor of rho-associated kinases
Ishizaki T, et al.
J. Mol. Pharmacol., 57, 976-983 (2000)

Articles

Naive pluripotent stem cells cultured in vitro using specialized media and inhibitors mimic "ground-state" cells from blastocysts.

Naive pluripotent stem cells cultured in vitro using specialized media and inhibitors mimic "ground-state" cells from blastocysts.

Naive pluripotent stem cells cultured in vitro using specialized media and inhibitors mimic "ground-state" cells from blastocysts.

Naive pluripotent stem cells cultured in vitro using specialized media and inhibitors mimic "ground-state" cells from blastocysts.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service