Skip to Content
Merck
All Photos(1)

Key Documents

L6293

Sigma-Aldrich

LY-364947

≥98% (HPLC)

Synonym(s):

4-[3-(2-pyridinyl)-1H-pyrazol-4-yl]-quinoline, HTS 466284, Transforming Growth Factor-β Type I Receptor Kinase Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H12N4
CAS Number:
Molecular Weight:
272.30
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

c1ccc(nc1)-c2n[nH]cc2-c3ccnc4ccccc34

InChI

1S/C17H12N4/c1-2-6-15-13(5-1)12(8-10-19-15)14-11-20-21-17(14)16-7-3-4-9-18-16/h1-11H,(H,20,21)

InChI key

IBCXZJCWDGCXQT-UHFFFAOYSA-N

Gene Information

human ... TGFBR1(7046)

Biochem/physiol Actions

LY-364947 is a selective, ATP-competitive inhibitor of Transforming Growth Factor-β Type I receptor kinase (TGF-β RI, ALK5) with IC50 = 59 nM. It is much less potent at related kinases, with IC50 = 400 nM for TGF-β RII and IC50 = 1400 nM for mixed lineage kinase-7 (MLK-7), a kinase in the MAP kinase signal pathway and closely related to TGF-β RII. LY-364947 inhibits TGF-β-dependent cellular growth (IC50 = 81 nM) in NIH 3T3 mouse fibroblasts.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Khaled Nassar et al.
Experimental eye research, 123, 72-86 (2014-04-20)
This study evaluates the use of the TGF-β receptor 1 inhibitor LY-364947 (LY) to prevent proliferative vitreoretinopathy (PVR). For the in vitro experiments Human Tenon's Fibroblasts (HTFs) and retinal pigment epithelial (RPE) cells were treated with different concentrations of LY
Suzanne Vanhauwaert et al.
PloS one, 9(10), e109091-e109091 (2014-10-14)
The selection and validation of stably expressed reference genes is a critical issue for proper RT-qPCR data normalization. In zebrafish expression studies, many commonly used reference genes are not generally applicable given their variability in expression levels under a variety
J Scott Sawyer et al.
Journal of medicinal chemistry, 46(19), 3953-3956 (2003-09-05)
Pyrazole-based inhibitors of the transforming growth factor-beta type I receptor kinase domain (TbetaR-I) are described. Examination of the SAR in both enzyme- and cell-based in vitro assays resulted in the emergence of two subseries featuring differing selectivity versus p38 MAP
Sheng-Ru Shiou et al.
The Journal of biological chemistry, 281(45), 33971-33981 (2006-09-09)
Metastasis is a primary cause of mortality due to cancer. Early metastatic growth involves both a remodeling of the extracellular matrix surrounding tumors and invasion of tumors across the basement membrane. Up-regulation of extracellular matrix degrading proteases such as urokinase
Hong-yu Li et al.
Journal of medicinal chemistry, 49(6), 2138-2142 (2006-03-17)
Novel dihydropyrrolopyrazole-substituted benzimidazoles were synthesized and evaluated in vitro as inhibitors of transforming growth factor-beta type I receptor (TGF-beta RI), TGF-beta RII, and mixed lineage kinase-7 (MLK-7). These compounds were found to be potent TGF-beta RI inhibitors and selective versus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service