Skip to Content
Merck
All Photos(1)

Key Documents

SML1961

Sigma-Aldrich

EF5

≥98% (HPLC)

Synonym(s):

2-(2-Nitro-1H-imidazol-1-yl)-N-(2,2,3,3,3-pentafluoropropyl)acetamide, EF-5, NSC 684681

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7F5N4O3
CAS Number:
Molecular Weight:
302.16
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NCC(F)(F)C(F)(F)F)CN1C=CN=C1[N+]([O-])=O

InChI

1S/C8H7F5N4O3/c9-7(10,8(11,12)13)4-15-5(18)3-16-2-1-14-6(16)17(19)20/h1-2H,3-4H2,(H,15,18)

InChI key

JGGDSDPOPRWSCX-UHFFFAOYSA-N

Biochem/physiol Actions

A 2-nitroimidazole-based hypoxia marker
EF5 is a 2-nitroimidazole-based hypoxia marker corresponding to a pentafluorinated derivative of the hypoxic cell-radiation sensitizer etanidazole. Under low oxygen (hypoxic) condition, reductive activation of its 2-nitroimidazole moiety leads to enhanced EF5 adduct formation with protein thiols and reduced glutathione. The pentafluorination derivatization substantially decreases the proportion of low molecular weight hydrolytic fragmentation of the adducts, making EF5 superior to etanidazole when utilized as a hypoxia marker by immunohistological staining.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E M Lord et al.
Cancer research, 53(23), 5721-5726 (1993-12-01)
Hypoxic cells in tissue pose many medical problems, and there is a need for more accurate measurements of tissue hypoxia. However, measurement of the pO2 and the extent of hypoxia within normal and tumor tissue have proven difficult. One of
Sung Hoon Cho et al.
Nature, 537(7619), 234-238 (2016-08-09)
Germinal centres (GCs) promote humoral immunity and vaccine efficacy. In GCs, antigen-activated B cells proliferate, express high-affinity antibodies, promote antibody class switching, and yield B cell memory. Whereas the cytokine milieu has long been known to regulate effector functions that
S M Evans et al.
British journal of cancer, 72(4), 875-882 (1995-10-01)
One of the most sensitive hypoxia detection methods is based on the observation that binding of nitroimidazoles to cellular macromolecules occurs as a result of hypoxia-dependent bioreduction by cellular nitroreductases. Nitroimidazole-binding techniques provide measurements of hypoxia to virtually any degree

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service