Skip to Content
Merck
All Photos(1)

Documents

N1165

Sigma-Aldrich

Pimonidazole

≥98% (HPLC)

Synonym(s):

NSC 380540, PD 126675;, Ro 03-8799;, alpha-((2-Nitroimidazol-1-yl)methyl)-1-piperidineethanol;, alpha-[(2-Nitro-1H-imidazol-1-yl)methyl]-1-piperidineethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H18N4O3
CAS Number:
Molecular Weight:
254.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

yellow

solubility

DMSO: >20 mg/mL

storage temp.

2-8°C

SMILES string

OC(CN1CCCCC1)Cn2ccnc2[N+]([O-])=O

InChI

1S/C11H18N4O3/c16-10(8-13-5-2-1-3-6-13)9-14-7-4-12-11(14)15(17)18/h4,7,10,16H,1-3,5-6,8-9H2

InChI key

WVWOOAYQYLJEFD-UHFFFAOYSA-N

Biochem/physiol Actions

Pimonidazole is an effective and nontoxic exogenous 2-nitroimidazole hypoxia marker. Pimonidazole forms adducts with thiol groups in proteins, peptides and amino acids.
Pimonidazole possesses a nitro group at position 2 and an imidazole ring. Pimonidazole undergoes reduction and is activated in hypoxic cells.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lena-Christin Conradi et al.
Angiogenesis, 20(4), 599-613 (2017-09-07)
Blockade of the glycolytic activator PFKFB3 in cancer cells (using a maximum tolerable dose of 70 mg/kg of the PFKFB3 blocker 3PO) inhibits tumor growth in preclinical models and is currently being tested as a novel anticancer treatment in phase I
Hypoxia Studies with Pimonidazole in vivo
Aguilera KY and Brekken RA
Bio-protocol, 4(19) (2014)
Yong Chen et al.
Disease models & mechanisms, 12(12) (2019-11-11)
Major risk factors for necrotizing enterocolitis (NEC) are formula feeding and prematurity; however, their pathogenic mechanisms are unknown. Here, we found that insufficient arginine/nitric oxide synthesis limits blood flow in the intestinal microvasculature, leading to hypoxia, mucosal damage and NEC
Pimonidazole: a novel hypoxia marker for complementary study of tumor hypoxia and cell proliferation in cervical carcinoma
Varia MA, et al.
Gynecologic Oncology, 71(2), 270-277 (1998)
Takahiro Minato et al.
Frontiers in physiology, 9, 478-478 (2018-06-06)
Fetal growth restriction (FGR) is a risk factor exacerbating a poor neurological prognosis at birth. A disease exacerbating a poor neurological prognosis is cerebral palsy. One of the cause of this disease is cerebral hemorrhage including intraventricular hemorrhage. It is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service