Skip to Content
Merck
All Photos(1)

Key Documents

SML1815

Sigma-Aldrich

Fasnall benzenesulfonate salt

≥98% (HPLC)

Synonym(s):

5,6-Dimethyl-N-[1-(phenylmethyl)-3-pyrrolidinyl]thieno[2,3-d]pyrimidin-4-amine benzenesulfonate, N-(1-Benzylpyrrolidin-3-yl)-5,6-dimethylthieno[2,3-d] pyrimidin-4-amine benzenesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22N4S · C6H5SO3H
CAS Number:
Molecular Weight:
496.64
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC(S1)=C(C)C2=C1N=CN=C2NC3CN(CC4=CC=CC=C4)CC3.O=S(C5=CC=CC=C5)(O)=O

InChI key

KLKWHGQIUKBXKT-UHFFFAOYSA-N

Application

Fasnall benzenesulfonate salt has been used as a fatty acid synthase inhibitor to test its effect on hypoxic U87 MG cells in cell viability assay and lipid droplet formation assay.

Biochem/physiol Actions

Fasnall is a fatty acid synthase (FASN or FAS) inhibitor (IC50 = 3.71 ?M by cell-free assay with 200 μM NADPH; IC50 = 147 and 213 nM against acetate and glucose lipids incorporation in HepG2 cells) composed of two enantiomers (HS-79 and HS-80) that selectively target FASN co-factor nucleotide-binding sites without affecting ACC, ZipK, AMPKα, AMPKγ, TRAP1, HSP70, NS5, IRAK2 nucleotide binding or the ATP-binding activity of BT474 cellular proteins. Fasnall is shown to completely block the proliferation in multiple breast cancer cultures at 50 μM as a result of apoptosis induction with much reduced cytotoxicity than C75 toward the non-tumorigenic cell line MCF10A. When administered via i.p. injection, Fasnall is efficacious in prolonging the survival of MMTV-Neu mice by effectively suppressing mammary adenocarcinoma tumor progression (15 mg/kg dosed alone twice weekly or 50 mg/kg dosed with carboplatin once weekly).
Fasnall is a fatty acid synthase (FASN or FAS) inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Raffaele Nicastro et al.
Nature cell biology, 25(9), 1303-1318 (2023-08-11)
Cell growth is regulated by the mammalian/mechanistic target of rapamycin complex 1 (mTORC1), which functions both as a nutrient sensor and a master controller of virtually all biosynthetic pathways. This ensures that cells are metabolically active only when conditions are
Anna Barkovskaya et al.
Molecular oncology, 15(8), 2026-2045 (2021-03-25)
Cellular phenotype plasticity between the epithelial and mesenchymal states has been linked to metastasis and heterogeneous responses to cancer therapy, and remains a challenge for the treatment of triple-negative breast cancer (TNBC). Here, we used isogenic human breast epithelial cell
Myriam Cerezo-Magaña et al.
Molecular cancer research : MCR, 19(3), 528-540 (2020-12-09)
As an adaptive response to hypoxic stress, aggressive tumors rewire their metabolic phenotype into increased malignant behavior through extracellular lipid scavenging and storage in lipid droplets (LD). However, the underlying mechanisms and potential lipid source retrieved in the hypoxic tumor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service