Skip to Content
Merck
All Photos(1)

Key Documents

N9876

Sigma-Aldrich

4-Nitrophenyl butyrate

lipase substrate, chromogenic, esterase substrate, ≥98%, liquid

Synonym(s):

p-Nitrophenyl butyrate, Butyric acid 4-nitrophenyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11NO4
CAS Number:
Molecular Weight:
209.20
Beilstein:
1879729
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Product Name

4-Nitrophenyl butyrate, ≥98%

Quality Level

Assay

≥98%

form

liquid

density

1.19 g/mL at 20 °C (lit.)

storage temp.

−20°C

SMILES string

CCCC(=O)Oc1ccc(cc1)[N+]([O-])=O

InChI

1S/C10H11NO4/c1-2-3-10(12)15-9-6-4-8(5-7-9)11(13)14/h4-7H,2-3H2,1H3

InChI key

DVDUMIQZEUTAGK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Nitrophenyl butyrate is a substrate for esterase, and lipase enzyme. Hydrolysis of 4-nitrophenyl butyrate by these enzymes releases the chromophore, 4-nitrophenolate which is spectrophotometrically analysed at 415 nm.

Application

4-nitrophenyl butyrate has been used:
  • as a substrate for esterase activity in monocyte-derived macrophage cell line in response to polyethylene
  • as a substrate for mycobacterial phospholipase A and mycobacterial cutinase activity
  • as a substrate to determine the effect of buffers and solvents on human carboxylesterase

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monitoring lipase/esterase activity by stopped flow in a sequential injection analysis system using p-nitrophenyl butyrate
Pliego J, et al.
Sensors, 15(2), 2798-2811 (2015)
Li-Chun Wang et al.
Journal of food and drug analysis, 26(3), 973-984 (2018-07-07)
Obesity is associated with higher risks of developing diabetes and cardiovascular disease. Green tea, rich in polyphenolic compounds such as epigallocatechin gallate (EGCG) and epigallocatechin (EGC), has been shown to display anti-obesity effects. Houttuynia cordata leaves have also been shown
Differential response to chemically altered polyethylene by activated mature human monocyte-derived macrophages
Xing S, et al.
Biomaterials, 23(17), 3595-3602 (2002)
Nathalia S Rios et al.
Biochimica et biophysica acta. Proteins and proteomics, 1867(9), 741-747 (2019-06-16)
The lipase from Pseudomonas fluorescens (PFL) has been immobilized on glyoxyl-octyl agarose and compared to the enzyme immobilized on octyl-agarose. Thus, PFL was immobilized at pH 7 on glyoxyl-octyl support via lipase interfacial activation and later incubated at pH 10.5 for 20 h
Geum Jin Kim et al.
Antioxidants (Basel, Switzerland), 10(2) (2021-02-10)
Since 2016, the invasive halophyte Spartina anglica has been colonizing mudflats along the western coast of South Korea. In order to minimize costs on S. anglica expansion management and waste-treatment of collected biomass, the potential application of the collected biomass

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service