Skip to Content
Merck
All Photos(2)

Key Documents

M7068

Sigma-Aldrich

MPP dihydrochloride hydrate

≥97% (HPLC), powder, estrogen receptor α agonist

Synonym(s):

1,3-Bis(4-hydroxyphenyl)-4-methyl-5-[4-(2-piperidinylethoxy)phenol]-1H-pyrazole dihydrochloride hydrate, Methyl-piperidino-pyrazole hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H31N3O3 · 2 HCl · xH2O
CAS Number:
Molecular Weight:
542.50 (anhydrous basis)
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

product name

MPP dihydrochloride hydrate, ≥97% (HPLC), white, powder

Quality Level

Assay

≥97% (HPLC)

form

powder

storage condition

desiccated

color

white

solubility

DMSO: soluble ≥20 mg/mL

SMILES string

Cl.Cl.Cc1c(nn(-c2ccc(O)cc2)c1-c3ccc(OCCN4CCCCC4)cc3)-c5ccc(O)cc5

InChI

1S/C29H31N3O3.2ClH/c1-21-28(22-5-11-25(33)12-6-22)30-32(24-9-13-26(34)14-10-24)29(21)23-7-15-27(16-8-23)35-20-19-31-17-3-2-4-18-31;;/h5-16,33-34H,2-4,17-20H2,1H3;2*1H

InChI key

FWDNPWVVRVSJQH-UHFFFAOYSA-N

General description

Methyl-piperidino-pyrazole hydrate (MPP) is prepared from a methyl-pyrazole-triol (MPT), a pyrazole triol.

Application

MPP dihydrochloride hydrate has been used as an estrogen receptor α antagonist:
  • in human breast cancer MCF-7 cells
  • to pre-treat endometrial ex vivo organ cultures (EVOCs)
  • to treat pituitary glands to monitor luteinizing hormone secretion

Biochem/physiol Actions

MPP dihydrochloride is a specific estrogen receptor α antagonist.
Methyl-piperidino-pyrazole hydrate (MPP) is an agonist for estrogen receptor α and lacks any ligand functionality with estrogen receptor (ER)β. It promotes silibinin induced apoptosis in human breast cancer MCF-7 cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nan Zheng et al.
Journal of pharmacological sciences, 128(3), 97-107 (2015-06-29)
The estrogen receptor alpha (ERα) has been proven to be one of the most important therapeutic targets in breast cancer over the last 30 years. Previous studies pointed out that a natural flavonoid, silibinin, induced apoptosis in human breast cancer
Fu Guangtao et al.
Frontiers in pharmacology, 12, 746391-746391 (2021-11-23)
Periprosthetic osteolysis is one of the major long-term complications following total joint replacement. Its cause is widely accepted to be wear particle-induced activation of inflammatory macrophages. No effective strategy for the prevention and treatment of periprosthetic osteolysis is yet available.
Julia Y H Liu et al.
General and comparative endocrinology, 272, 63-75 (2018-12-06)
Estrogen is well known to have a modulatory role on gastrointestinal tract, particularly through its interaction with nuclear estrogen receptors (ERs), alpha and beta (ERα/β). Recent functional studies also indicate that estrogen can activate a G-protein coupled estrogen receptor, GPR30
Hai-Bing Zhou et al.
Bioorganic & medicinal chemistry letters, 19(1), 108-110 (2008-11-19)
Methyl-piperidino-pyrazole (MPP), an estrogen receptor alpha (ERalpha)-selective antagonist we developed, has a basic side chain (BSC) attached to an ERalpha-selective agonist ligand, methyl-pyrazole-triol (MPT) through an ether linkage. To remove the possibility that metabolic cleavage of the BSC in MPP
João P E Saut et al.
Reproduction (Cambridge, England), 148(6), 593-606 (2014-09-24)
The risk of bacterial infection of the endometrium causing uterine disease in cattle is increased in the progesterone-dominated luteal phase of the ovarian cycle, while oestrogens or oestrus are therapeutic or protective against disease. The first line of defence against

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service