Skip to Content
Merck
All Photos(1)

Key Documents

SML1682

Sigma-Aldrich

CDN1163

≥98% (HPLC)

Synonym(s):

4-(1-Methylethoxy)-N-(2-methyl-8-quinolinyl)-benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H20N2O2
CAS Number:
Molecular Weight:
320.39
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CC1=NC2=C(NC(C3=CC=C(OC(C)C)C=C3)=O)C=CC=C2C=C1

InChI

1S/C20H20N2O2/c1-13(2)24-17-11-9-16(10-12-17)20(23)22-18-6-4-5-15-8-7-14(3)21-19(15)18/h4-13H,1-3H3,(H,22,23)

InChI key

GVGVYDCVFBGALZ-UHFFFAOYSA-N

Biochem/physiol Actions

CDN1163 is an allosteric activator of sarco/ endoplasmic reticulum Ca2+-ATPase (SERCA) that potently lowered fasting blood glucose, improved glucose tolerance and ameliorated hepatosteatosis in ob/ob mice. Additionally CDN1163 induces significant reduction in adipose tissue weight with no change in lean mass in ob/ob mice. CDN1163 has no effect on glucose levels or body weight in lean control mice.
CDN1163 treatment declined the hepatic expression of genes implicated in gluconeogenesis and lipogenesis as well as attenuated endoplasmic reticulum (ER) stress response and ER stress-induced apoptosis. CDN1163 interacts with sarco/endoplasmic reticulum Ca2+-ATPase (SERCA) and enhances ER Ca2+ content.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Russell Dahl
Bioorganic & medicinal chemistry, 25(1), 53-57 (2016-10-26)
Endoplasmic reticulum (ER) stress is intimately linked to Parkinson's disease (PD) pathophysiology. Disrupted intracellular calcium homeostasis is a major cause of the ER stress seen in dopaminergic neurons, leading to the cell death and subsequent loss of movement and coordination
Katherine Krajnak et al.
Bioorganic & medicinal chemistry letters, 28(9), 1591-1594 (2018-04-01)
Amongst the cellular cacophony of altered signals in Alzheimer's disease (AD), disrupted Ca2+ homeostasis and consequential endoplasmic reticulum (ER) stress signals have been recognized as key determinants of neuron fate. This altered Ca2+ state is accompanied by a failing sarco/endoplasmic
Cytoprotective small molecule modulators of endoplasmic reticulum stress.
Munshi S and Russell D
Bioorganic & Medicinal Chemistry, 24(11), 2382-2388 (2016)
Small molecular allosteric activator of the sarco/endoplasmic reticulum Ca2+-ATPase (SERCA) attenuates diabetes and metabolic disorders.
Kang S, et al.
The Journal of Biological Chemistry, 391(10), 5185-5198 (2016)
M'Lynn E Fisher et al.
eLife, 10 (2021-06-03)
The sarco-plasmic reticulum calcium pump (SERCA) plays a critical role in the contraction-relaxation cycle of muscle. In cardiac muscle, SERCA is regulated by the inhibitor phospholamban. A new regulator, dwarf open reading frame (DWORF), has been reported to displace phospholamban

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service