Skip to Content
Merck
All Photos(1)

Key Documents

C7802

Sigma-Aldrich

Charybdotoxin

≥90% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C176H277N57O55S7
CAS Number:
Molecular Weight:
4295.89
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32
Pricing and availability is not currently available.

Quality Level

Assay

≥90% (HPLC)

form

powder

storage temp.

−20°C

SMILES string

S1SC[C@@H]2NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@H](CSSC[C@@H]5NC(=O)[C@@H](NC(=O)CNC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)

InChI

1S/C176H277N57O55S7/c1-81(2)58-106-149(263)213-110(62-91-67-191-80-197-91)152(266)215-112(64-127(183)246)156(270)231-135(85(8)240)170(284)219-114(69-234)157(271)200-95(36-25-54-192-174(185)186)138(252)196-68-130(249)199-96(32-17-21-50-177)139(253)222-120-75-291-295-79-124(226-159(273)116(71-236)218-167(281)132(82(3)4)228-155(269)113(65-128(184)247)216-169(283)134(84(7)239)230-154(268)108(59-88-28-13-12-14-29-88)210-145(259)102-44-47-129(248)198-102)166(280)232-137(87(10)242)172(286)233-136(86(9)241)171(285)220-115(70-235)158(272)204-98(34-19-23-52-179)141(255)206-104(45-48-131(250)251)147(261)225-122-77-292-290-74-119(223-143(257)99(35-20-24-53-180)201-140(254)97(33-18-22-51-178)203-153(267)111(63-126(182)245)214-148(262)105(49-57-289-11)208-162(120)276)161(275)205-101(38-27-56-194-176(189)190)144(258)224-121(164(278)211-107(60-89-39-41-92(243)42-40-89)150(264)221-118(73-238)173(287)288)76-293-294-78-123(163(277)207-103(43-46-125(181)244)146(260)202-100(142(256)209-106)37-26-55-193-175(187)188)227-168(282)133(83(5)6)229-160(274)117(72-237)217-151(265)109(212-165(122)279)61-90-66-195-94-31-16-15-30-93(90)94/h12-16,28-31,39-42,66-67,80-87,95-124,132-137,195,234-243H,17-27,32-38,43-65,68-79,177-180H2,1-11H3,(H2,181,244)(H2,182,245)(H2,183,246)(H2,184,247)(H,191,197)(H,196,252)(H,198,248)(H,199,249)(H,200,271)(H,201,254)(H,202,260)(H,203,267)(H,204,272)(H,205,275)(H,206,255)(H,207,277)(H,208,276)(H,209,256)(H,210,259)(H,211,278)(H,212,279)(H,213,263)(H,214,262)(H,215,266)(H,216,283)(H,217,265)(H,218,281)(H,219,284)(H,220,285)(H,221,264)(H,222,253)(H,223,257)(H,224,258)(H,225,261)(H,226,273)(H,227,282)(H,228,269)(H,229,274)(H,230,268)(H,231,270)(H,232,280)(H,233,286)(H,250,251)(H,287,288)(H4,185,186,192)(H4,187,188,193)(H4,189,190,194)/t84-,85-,86-,87-,95+,96+,97+,98+,99+,100+,101+,102+,103+,104+,105+,106+,107+,108+,109+,110+,111+,112+,113+,114+,115+,116+,117+,118+,119+,120+,121+,122+,123+,124+,132+,133+,134+,135+,136+,137+/m1/s1

InChI key

CNVQLPPZGABUCM-LIGYZCPXSA-N

Gene Information

human ... KCNA3(3738)
mouse ... KCNA3(16491)
rat ... KCNA3(29731)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Glp-Phe-Thr-Asn-Val-Ser-Cys-Thr-Thr-Ser-Lys-Glu-Cys-Trp-Ser-Val-Cys-Gln-Arg-Leu-His-Asn-Thr-Ser-Arg-Gly-Lys-Cys-Met-Asn-Lys-Lys-Cys-Arg-Cys-Tyr-Ser [Disulfide Bridges: 7-28, 13-33, 17-35]

Application

Charybdotoxin has been used:
  • in the inhibition of endothelium-derived hyperpolarizing factor responses[1]
  • in electrophysiological studies, as calcium channel blockers[2][3]
  • as a transport inhibitor[4]

Biochem/physiol Actions

Charybdotoxin (ChTX) is a scorpion venom, which blocks a voltage-gated potassium channel,[3] by binding to the extracellular loop of the potassium channel.[5]
High (BK) or intermediate (IK1) conductance calcium-activated and voltage-gated (Kv1.3) channel blocker.
Kv1.3 channel blocker.
Potent and selective inhibitor of the Ca2+-activated K+ channel present in GH3 anterior pituitary cells and primary bovine aortic smooth muscle cells.

Other Notes

Peptide found in the venom of the scorpion Leiurus quinquestriatus.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

J M Doughty et al.
The American journal of physiology, 276(3 Pt 2), H1107-H1112 (1999-03-10)
In rat mesenteric artery, endothelium-derived hyperpolarizing factor (EDHF) is blocked by a combination of apamin and charybdotoxin (ChTX). The site of action of these toxins has not been established. We compared the effects of ChTX and apamin applied selectively to
Transepithelial impedance analysis of chloride secretion.
Ashvani K Singh et al.
Methods in molecular medicine, 70, 129-142 (2002-03-29)
Cultured heart cells from oyster: an experimental approach for evaluation of the toxicity of the marine pollutant tributyltin
Droguet M, et al.
Aquatic Living Resources, 25(2), 185-194 (2012)
Solution structure of extracellular loop of human beta4 subunit of BK channel and its biological implication on ChTX sensitivity
Wang Y, et al.
Scientific Reports, 8(1), 4571-4571 (2018)
The charybdotoxin family of K+ channel-blocking peptides.
C Miller
Neuron, 15(1), 5-10 (1995-07-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service