Skip to Content
Merck
All Photos(4)

Key Documents

P9767

Sigma-Aldrich

Sodium palmitate

≥98.5%

Synonym(s):

Hexadecanoic acid sodium salt, Palmitic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)14COONa
CAS Number:
Molecular Weight:
278.41
Beilstein:
3575882
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

plant (palm)

Assay

≥98.5%

form

powder

mp

283-290 °C (lit.)

functional group

ester

lipid type

saturated FAs

shipped in

ambient

storage temp.

2-8°C

SMILES string

[Na+].CCCCCCCCCCCCCCCC([O-])=O

InChI

1S/C16H32O2.Na/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18;/h2-15H2,1H3,(H,17,18);/q;+1/p-1

InChI key

GGXKEBACDBNFAF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium palmitate (PA) has been used:
  • to induce inflammation and thrombosis pathway in murine macrophage cell line RAW 264.7 cell line by activating reactive oxygen species (ROS) production, Janus-kinase (JNK) signalling and release of histone H3 by western blotting and cell viability by MTT assay
  • to induce lipogenesis in AML12 cells and primary hepatocytes to analyse the effect of irisin on PA induced lipogenesis and related signal pathways by western blot analysis and quantitative PCR analysis
  • as a component in free fatty acid mixture to induce cellular steatosis in HepG2 cell lines and determination of lipid accumulation by Oil-Red-O staining

Biochem/physiol Actions

Sodium palmitate is the sodium salt of palmitic acid, a component in hard soaps. Palmitic acid is a common saturated fatty acid and produced during fatty acid synthesis. Sodium palmitate enhances lipogenesis, cellular steatosis in various cell lines. Palmitate induces cell death in human epidermal growth factor receptor 2 (HER2)/neu-positive cells and breast cancer cell lines like MCF-7 due to enhanced fatty acid accumulation. Sodium palmitate induces lipoapoptosis in L02 and HepG2 liver cells by inducing glycogen synthase kinase-3β (GSK3β) expression.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

José M S Fernández-Calleja et al.
Scientific reports, 9(1), 11507-11507 (2019-08-10)
Indirect calorimetry (InCa) estimates whole-body energy expenditure and total substrate oxidation based on O2 consumption and CO2 production, but does not allow for the quantification of oxidation of exogenous substrates with time. To achieve this, we incorporated 13CO2 and 12CO2
Metabolic assays for detection of neutral fat stores
Baumann JM, et al.
Bio-protocol, 90(5), 20-20 (2015)
Saturated fatty acid palmitate induces extracellular release of histone H3: a possible mechanistic basis for high-fat diet-induced inflammation and thrombosis
Shrestha C, et al.
Biochemical and Biophysical Research Communications, 437(4), 573-578 (2013)
Kyong Kim et al.
Nutrients, 11(7) (2019-07-07)
Allomyrina dichotoma larva is a nutritional-worthy future food resource and it contributes to multiple pharmacological functions. However, its antidiabetic effect and molecular mechanisms are not yet fully understood. Therefore, we investigated the hypolipidemic effect of A. dichotoma larva extract (ADLE)
New role of irisin in hepatocytes: The protective effect of hepatic steatosis in vitro
Park MJ, et al.
Cellular Signalling, 27(9), 1831-1839 (2015)

Articles

Fatty acid synthesis supports cancer cell proliferation, essential for membrane generation, protein modification, and bioenergetics.

Fatty acid synthesis supports cancer cell proliferation, essential for membrane generation, protein modification, and bioenergetics.

Fatty acid synthesis supports cancer cell proliferation, essential for membrane generation, protein modification, and bioenergetics.

Fatty acid synthesis supports cancer cell proliferation, essential for membrane generation, protein modification, and bioenergetics.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service