Skip to Content
Merck
All Photos(1)

Key Documents

PZ0013

Sigma-Aldrich

Nelfinavir mesylate hydrate

≥98% (HPLC)

Synonym(s):

3S,4aS,8aS)-N-(1,1-Dimethylethyl)decahydro-2-[(2R,3R)-2-hydroxy-3-[(3-hydroxy-2-methylbenzoyl)amino]-4-(phenylthio)butyl]-3-isoquinolinecarboxamide methanesulfonate, AG-1343, Nelfinavir methanesulfonate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H45N3O4S · CH3SO3H · xH2O
CAS Number:
Molecular Weight:
663.89 (anhydrous basis)
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: ≥2 mg/mL

storage temp.

2-8°C

SMILES string

CC1=C(O)C=CC=C1C(N[C@@H](CSC2=CC=CC=C2)[C@H](O)CN3[C@H](C(NC(C)(C)C)=O)C[C@@]4([H])[C@@](CCCC4)([H])C3)=O.[CH2]S(O)(=O)=O

InChI

1S/C32H45N3O4S.CH4O3S/c1-21-25(15-10-16-28(21)36)30(38)33-26(20-40-24-13-6-5-7-14-24)29(37)19-35-18-23-12-9-8-11-22(23)17-27(35)31(39)34-32(2,3)4;1-5(2,3)4/h5-7,10,13-16,22-23,26-27,29,36-37H,8-9,11-12,17-20H2,1-4H3,(H,33,38)(H,34,39);1H3,(H,2,3,4)/t22-,23+,26-,27-,29+;/m0./s1

InChI key

NQHXCOAXSHGTIA-SKXNDZRYSA-N

Related Categories

Application

Nelfinavir mesylate hydrate has been used to test its therapeutic effect on pulmonary fibrosis in patients with systemic sclerosis (SSc). It has also been used in combination with bortezomib to selectively kill tuberous sclerosis complex 2 (TSC2)-deficient tumor cells.

Biochem/physiol Actions

Nelfinavir is a HIV protease inhibitor; antiretroviral; and anti-tumor agent.
Nelfinavir mesylate hydrate, also known as AG1343, is used as a therapeutic agent for acquired immunodeficiency syndrome (AIDS). It acts as an antifibrotic agent and is used to treat pulmonary fibrosis in patients with systemic sclerosis (SSc). In addition, nelfinavir mesylate hydrate reduces inflammation by activating protein phosphatase 2 (PP-2) and blocking mitogen-activated protein kinases (MAPK) signaling in macrophages. It also stimulate autophagy and exhibit anti-cancer property.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karin Nielsen-Saines et al.
The New England journal of medicine, 366(25), 2368-2379 (2012-06-22)
The safety and efficacy of adding antiretroviral drugs to standard zidovudine prophylaxis in infants of mothers with human immunodeficiency virus (HIV) infection who did not receive antenatal antiretroviral therapy (ART) because of late identification are unclear. We evaluated three ART
Joong Sup Shim et al.
Journal of the National Cancer Institute, 104(20), 1576-1590 (2012-10-09)
Human epidermal growth factor receptor 2 (HER2)-positive breast cancer is highly aggressive and has higher risk of recurrence than HER2-negative cancer. With few treatment options available, new drug targets specific for HER2-positive breast cancer are needed. We conducted a pharmacological
Simmy Thomas et al.
Cancer letters, 325(1), 63-71 (2012-06-06)
The cellular processes of autophagy and endoplasmic reticulum stress (ERS) appear to be interconnected, and it has been proposed that autophagy may serve to reduce ERS via removal of terminally misfolded and aggregated proteins. Conversely, there are indications that blockage
Adam M Petrich et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 18(9), 2534-2544 (2012-02-18)
The mTOR pathway is constitutively activated in diffuse large B-cell lymphoma (DLBCL). mTOR inhibitors have activity in DLBCL, although response rates remain low. We evaluated DLBCL cell lines with differential resistance to the mTOR inhibitor rapamycin: (i) to identify gene
The Antiretroviral Agent Nelfinavir Mesylate: A Potential Therapy for Systemic Sclerosis
Sanchez C, et al.
Arthritis & Rheumatology (Hoboken, N.J.), 70(1), 115-126 (2018)

Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service