SML1169
Pitstop 2
≥98% (HPLC), powder, clathrin-mediated endocytosis inhibitor
Synonym(s):
N-[5-[(4-Bromophenyl)methylene]-4,5-dihydro-4-oxo-2-thiazolyl]-1-naphthalenesulfonamide, Pitstop-2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C20H13BrN2O3S2
CAS Number:
Molecular Weight:
473.36
UNSPSC Code:
12352200
NACRES:
NA.77
Recommended Products
Product Name
Pitstop 2, ≥98% (HPLC)
Quality Level
Assay
≥98% (HPLC)
form
powder
color
white to light brown
solubility
DMSO: 20 mg/mL, clear
storage temp.
−20°C
Application
Pitstop 2 has been used as an inhibitor of clathrin-mediated endocytosis.
Biochem/physiol Actions
Pitstop 2 also has an ability to breakdown the nuclear pore complex (NPC) permeability barrier and decrease importin β binding along with alteration of the NPC ultrastructure. Pitstop 2 is also involved in the inhibition of platelet activation, platelet aggregation and secretion.
Pitstop 2 is a selective inhibitor of clathrin-mediated endocytosis that acts via blocking ligand access to the clathrin terminal domain. Pitstop 2 selectively inhibits clathrin-mediated HIV entry and stalls clathrin-coated pit (CCP) dynamics.
Pitstop 2 is a selective inhibitor of clathrin-mediated endocytosis.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Clathrin inhibitor Pitstop-2 disrupts the nuclear pore complex permeability barrier
Liashkovich I, et al.
Scientific Reports, 5, 9994-9994 (2015)
Clathrin-mediated integrin ?IIb?3 trafficking controls platelet spreading
Gao W, et al.
Platelets, 1-12 (2017)
Muhammad Asam Riaz et al.
Journal of proteomics, 227, 103918-103918 (2020-07-28)
Bacterial insecticidal proteins, such as the Bin toxin from Lysinibacillus sphaericus, could be used more extensively to control insecticide resistant mosquitoes. This study was aimed at identification of mosquito cell proteins binding Bin toxin. Results showed that purified toxin was
Leucine-rich repeat-containing G protein?coupled receptor 4 facilitates vesicular stomatitis virus infection by binding vesicular stomatitis virus glycoprotein
Zhang N, et al,
The Journal of Biological Chemistry, 292(40), 16527-16538 (2017)
Meagan M Postema et al.
Molecular biology of the cell, 30(19), 2515-2526 (2019-08-08)
Apical microvilli are critical for the homeostasis of transporting epithelia, yet mechanisms that control the assembly and morphology of these protrusions remain poorly understood. Previous studies in intestinal epithelial cell lines suggested a role for the F-BAR domain protein PACSIN2
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service