Skip to Content
Merck
All Photos(2)

Documents

A3085

Sigma-Aldrich

5-(N-Ethyl-N-isopropyl)amiloride

≥98% (TLC), powder, Na⁺/H⁺ antiport inhibitor

Synonym(s):

EIPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H18ClN7O
CAS Number:
Molecular Weight:
299.76
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

5-(N-Ethyl-N-isopropyl)amiloride,

biological source

synthetic

Quality Level

Assay

≥98% (TLC)

form

powder

mp

202-205 °C (lit.)

solubility

methanol: 9.80-10.20 mg/mL, clear, light yellow to yellow

storage temp.

2-8°C

SMILES string

CCN(C(C)C)c1nc(N)c(nc1Cl)C(=O)NC(N)=N

InChI

1S/C11H18ClN7O/c1-4-19(5(2)3)9-7(12)16-6(8(13)17-9)10(20)18-11(14)15/h5H,4H2,1-3H3,(H2,13,17)(H4,14,15,18,20)

InChI key

QDERNBXNXJCIQK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5-(N-Ethyl-N-isopropyl)amiloride has been used:
  • as a supplement in endothelial cell growth medium to incubate human lung microvascular endothelial cell
  • in Pluronic F-127 stock solution for Drosophila larval live-imaging
  • in choline chloride solution as an experimental perfusate, to study the effect of Na+/H+ inhibition on jejunal loop perfusion in vivo

Biochem/physiol Actions

Selective blocker of Na+/H+ antiport

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhao Luo et al.
Journal of peptide science : an official publication of the European Peptide Society, 22(11-12), 689-699 (2016-10-16)
Cell-penetrating peptides (CPPs) have been shown to be potential drug carriers for cancer therapy. The inherently low immunogenicity and cytotoxicity of human-derived CPPs make them more suitable for intracellular drug delivery compared to other delivery vehicles. In this work, the
Rachael L Hardison et al.
mSphere, 3(5) (2018-09-14)
Nutrient limitation restricts bacterial growth in privileged sites such as the middle ear. Transient heme-iron restriction of nontypeable Haemophilus influenzae (NTHI), the major causative agent of chronic and recurrent otitis media (OM), promotes new and diverse phenotypes that can influence
An investigation into the relationship between small intestinal fluid secretion and systemic arterial blood pressure in the anesthetized rat
Lucas ML and Morrison JD
Physiological Reports, 3(5) (2015)
Xinquan Liu et al.
International journal of nanomedicine, 14, 6589-6600 (2019-09-10)
The RAS family of oncogenes (KRAS, HRAS, NRAS) are the most frequent mutations in cancers and regulate key signaling pathways that drive tumor progression. As a result, drug delivery targeting RAS-driven tumors has been a long-standing challenge in cancer therapy.
Dipali Patel et al.
Scientific reports, 7(1), 12886-12886 (2017-10-12)
Mammalian mitochondria can be transferred between cells both in culture and in vivo. There is evidence that isolated mitochondria enter cells by endocytosis, but the mechanism has not been fully characterised. We investigated the entry mechanism of isolated mitochondria into

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service