Skip to Content
Merck
All Photos(1)

Key Documents

SML0407

Sigma-Aldrich

ML 141

≥98% (HPLC)

Synonym(s):

4-[4,5-Dihydro-5-(4-methoxyphenyl)-3-phenyl-1H-pyrazol-1-yl]-benzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H21N3O3S
CAS Number:
Molecular Weight:
407.49
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL (warmed, clear solution)

storage temp.

2-8°C

SMILES string

NS(C1=CC=C(C=C1)N2N=C(C3=CC=CC=C3)CC2C4=CC=C(OC)C=C4)(=O)=O

InChI

1S/C22H21N3O3S/c1-28-19-11-7-17(8-12-19)22-15-21(16-5-3-2-4-6-16)24-25(22)18-9-13-20(14-10-18)29(23,26)27/h2-14,22H,15H2,1H3,(H2,23,26,27)

InChI key

QBNZBMVRFYREHK-UHFFFAOYSA-N

Related Categories

Application

ML 141 has been used:
  • to inhibit CDC42 GTPase in human immortalized gingival epithelial (HIGE) cells
  • as inhibitors of Rho kinase to study the role of small Rho GTPases on localization of peripheral nuclei
  • as actin regulator inhibitor, to determine which actin regulators and nucleators are involved in the assembly of F-actin cages around damaged mitochondria
  • as a selective, non-competitive inhibitor of Cdc42 to treat CCD-1070Sk cells

Biochem/physiol Actions

ML 141 is a potent, selective inhibitor of the Rho family GTPase cdc42.
ML 141 is a potent, selective inhibitor of the Rho family GTPase cdc42. The IC50 for inhibition of enzymatic activity is 200 nM, with no activity against Rho family members Rac, Ras or Rab. Cdc42 has been implicated in the regulation of actin polymerization through its direct binding to Neural Wiskott-Aldrich syndrome protein (N-WASP), which subsequently activates Arp2/3 complex. This complex mediates the polymerization of actin into branched networks and regulates important functions including cell adhesion, cytoskeletal arrangement, phagocytosis and host-pathogen interactions, motility, migration, and membrane protein trafficking.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the GTP Binding Proteins (Low Molecular Weight) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intracellular periodontal pathogen exploits recycling pathway to exit from infected cells
Takeuchi H, et al.
Cellular Microbiology, 18(7), 928-948 (2016)
Multiple routes of endocytic internalization of PDGFR beta contribute to PDGF-induced STAT3 signaling
Jastrzkebski K, et al.
Journal of Cell Science, 130(3), 577-589 (2017)
Local arrangement of fibronectin by myofibroblasts governs peripheral nuclear positioning in muscle cells
Roman W, et al.
Developmental Cell, 46(1), 102-111 (2018)
Brennan D Gerlach et al.
Cell death and differentiation, 27(2), 525-539 (2019-06-22)
Inflammation-resolution is a protective response that is mediated by specialized pro-resolving mediators (SPMs). The clearance of dead cells or efferocytosis is a critical cellular program of inflammation-resolution. Impaired efferocytosis can lead to tissue damage in prevalent human diseases, like atherosclerosis.
Benjamin Ledoux et al.
Science advances, 9(50), eade1660-eade1660 (2023-12-13)
In their environment, cells must cope with mechanical stresses constantly. Among these, nanoscale deformations of plasma membrane induced by substrate nanotopography are now largely accepted as a biophysical stimulus influencing cell behavior and function. However, the mechanotransduction cascades involved and

Articles

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service