Skip to Content
Merck
All Photos(1)

Key Documents

SML0006

Sigma-Aldrich

CK-666

≥98% (HPLC), powder, Arp2/3 comple inhibitor

Synonym(s):

2-Fluoro-N-[2-(2-methyl-1H-indol-3-yl)ethyl]-benzamide, CK 666, CK-0944666

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H17FN2O
CAS Number:
Molecular Weight:
296.34
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

CK-666, ≥98% (HPLC), powder

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥25 mg/mL

storage temp.

2-8°C

SMILES string

Cc1[nH]c2ccccc2c1CCNC(=O)c3ccccc3F

InChI

1S/C18H17FN2O/c1-12-13(14-6-3-5-9-17(14)21-12)10-11-20-18(22)15-7-2-4-8-16(15)19/h2-9,21H,10-11H2,1H3,(H,20,22)

InChI key

UXRKUKRXVWJFER-UHFFFAOYSA-N

Application

CK-666 may be used to study Arp2/3-mediated structural changes in cells.

Biochem/physiol Actions

CK-666 binds to Arp2/3 complex, stabilizes the inactive state of the complex and prevents its movement into active conformation.
CK-666 is a cell-permeable inhibitor of actin assembly mediated by actin-related protein Arp2/3 complex.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Byron Hetrick et al.
Chemistry & biology, 20(5), 701-712 (2013-04-30)
Actin-related protein 2/3 (Arp2/3) complex is a seven-subunit assembly that nucleates branched actin filaments. Small molecule inhibitors CK-666 and CK-869 bind to Arp2/3 complex and inhibit nucleation, but their modes of action are unknown. Here, we use biochemical and structural
Akriti Prashar et al.
Frontiers in cellular and infection microbiology, 8, 133-133 (2018-05-19)
Legionella pneumophila (Lp) exhibits different morphologies with varying degrees of virulence. Despite their detection in environmental sources of outbreaks and in respiratory tract secretions and lung autopsies from patients, the filamentous morphotype of Lp remains poorly studied. We previously demonstrated
Aaron S Dhanda et al.
mBio, 9(2) (2018-04-11)
Palladin is an important component of motile actin-rich structures and nucleates branched actin filament arrays in vitro Here we examine the role of palladin during Listeria monocytogenes infections in order to tease out novel functions of palladin. We show that
Satoko Sakamoto et al.
PLoS biology, 16(9), e2004874-e2004874 (2018-09-27)
Formin is one of the two major classes of actin binding proteins (ABPs) with nucleation and polymerization activity. However, despite advances in our understanding of its biochemical activity, whether and how formins generate specific architecture of the actin cytoskeleton and
Yuki M Shirai et al.
PloS one, 12(11), e0188778-e0188778 (2017-12-01)
Electron tomography of the plasma membrane (PM) identified several layers of cortical actin meshwork running parallel to the PM cytoplasmic surface throughout the PM. Here, cortical actin structures and dynamics were examined in living cells, using super-resolution microscopy, with (x,y)-

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service