Skip to Content
Merck
All Photos(1)

Key Documents

SML0292

Sigma-Aldrich

Gestodene

≥98% (HPLC)

Synonym(s):

(17α)-13-Ethyl-17-hydroxy-18,19-Dinorpregna-4,15-dien-20-yn-3-one, Gestinol, SHB 331, WL 70

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26O2
CAS Number:
Molecular Weight:
310.43
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -175 to -195° (DCM)

color

white to off-white

solubility

DMSO: >20 mg/mL

storage temp.

−20°C

SMILES string

CC[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@H]34)[C@@H]1C=C[C@@]2(O)C#C

InChI

1S/C21H26O2/c1-3-20-11-9-17-16-8-6-15(22)13-14(16)5-7-18(17)19(20)10-12-21(20,23)4-2/h2,10,12-13,16-19,23H,3,5-9,11H2,1H3/t16-,17+,18+,19-,20-,21-/m0/s1

InChI key

SIGSPDASOTUPFS-XUDSTZEESA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Gestodene is a synthetic progestin used as a contraceptive. Gestodene displays a high binding affinity to the progesterone receptor, and also binds strongly to adrogen and glucocorticoid receptors.

Features and Benefits

This compound is featured on the Nuclear Receptors (Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pasquale Florio et al.
Gynecological endocrinology : the official journal of the International Society of Gynecological Endocrinology, 27(6), 434-438 (2011-01-06)
In a retrospective case-control study, we compared the effectiveness of hysteroscopic correction and hormonal treatment to improve symptoms [postmestrual abnormal uterine bleeding (PAUB), pelvic pain localized in suprapubic site] associated with isthmocele. Women (n = 39; mean age ± SD
Ana Cristina Rebelo et al.
The European journal of contraception & reproductive health care : the official journal of the European Society of Contraception, 16(4), 289-297 (2011-07-22)
To evaluate the influence of oral contraceptives (OCs) containing 20 μg ethinylestradiol (EE) and 150 μg gestodene (GEST) on the autonomic modulation of heart rate (HR) in women. One-hundred and fifty-five women aged 24 ± 2 years were divided into
Combined oral contraceptives, venous thromboembolism, and the problem of interpreting large but incomplete datasets.
Jürgen Dinger et al.
The journal of family planning and reproductive health care, 38(1), 2-6 (2011-12-01)
Hormone-based contraceptive therapy and risk of venous thromboembolism in young women.
Helen Roberts
Clinical advances in hematology & oncology : H&O, 8(5), 307-309 (2010-06-17)
Ana Cristina S Rebelo et al.
Contraception, 81(4), 309-315 (2010-03-17)
The purpose of this study was to evaluate the effect of long-term use of oral contraceptives (OC) containing 0.20 mg of ethinylestradiol (EE) combined with 0.15 mg of gestodene (GEST) on the peak aerobic capacity and at the anaerobic threshold

Articles

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service