Skip to Content
Merck
All Photos(3)

Key Documents

E4876

Sigma-Aldrich

17α-Ethynylestradiol

≥98%

Synonym(s):

17α-Ethynyl-1,3,5(10)-estratriene-3,17β-diol, 19-Nor-1,3,5(10),17α-pregnatrien-20-yne-3,17-diol, Ethinylestradiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24O2
CAS Number:
Molecular Weight:
296.40
Beilstein:
2419975
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

biological source

synthetic

sterility

non-sterile

Assay

≥98%

form

powder

mp

182-183 °C (lit.)

solubility

ethanol: 50 mg/mL, clear, colorless to faintly yellow

shipped in

ambient

storage temp.

room temp

SMILES string

[H][C@]12CC[C@@]3(C)[C@@]([H])(CC[C@@]3(O)C#C)[C@]1([H])CCc4cc(O)ccc24

InChI

1S/C20H24O2/c1-3-20(22)11-9-18-17-6-4-13-12-14(21)5-7-15(13)16(17)8-10-19(18,20)2/h1,5,7,12,16-18,21-22H,4,6,8-11H2,2H3/t16-,17-,18+,19+,20+/m1/s1

InChI key

BFPYWIDHMRZLRN-SLHNCBLASA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

17α-Ethynylestradiol (EE2) is a synthetic hormone. It is a derivative of estradiol (E2).[1] It is implicated in estrogen replacement therapy and suspension of breastfeeding.[2] EE2 is used to treat osteoporosis, menopausal and postmenopausal syndrome. It is also used to treat prostatic cancer and breast cancer in postmenopausal women.[1] EE2 functions as an endocrine-disrupting chemical (EDC).[2]

Application

17α-Ethynylestradiol has been used to study the effect of exogenous estrogen and selective estrogen receptor modulator (SERM) in rats.[3] It also has been used to increase the concentration of low density lipoprotein (LDL) receptor in rabbit liver.[4]

Biochem/physiol Actions

17α-Ethynylestradiol is an orally bio-active synthetic estrogen used as an oral contraceptive.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 1 - Carc. 2 - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of 17-beta-estradiol and 17-alpha-ethinylestradiol in biological and environmental matrices?a review
Barreiros L, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 126, 243-262 (2016)
Purification of low density lipoprotein receptor from liver and its quantification by anti-receptor monoclonal antibodies
Gherardi E, et al.
The Biochemical Journal, 253(2), 409-415 (1988)
Huda Omar Ali et al.
PloS one, 11(3), e0152114-e0152114 (2016-03-22)
Oestrogens influence the pathology and development of hormone-sensitive breast cancers. Tissue factor pathway inhibitor (TFPI) has been shown to be associated with breast cancer pathogenesis. Recently, we found TFPI mRNA levels to be significantly reduced by oestrogens in a breast
Occurrence of 17alpha-ethynylestradiol (EE2) in the environment and effect on exposed biota: a review
Aris AZ, et al.
Environment International, 69, 104-119 (2014)
Ovariectomized rats as a model of postmenopausal osteoarthritis: validation and application
Hoegh-Andersen P, et al.
Arthritis Research & Therapy, 6(2), R169-R169 (2004)

Questions

  1. 17α-Ethynylestradiol 물질이 mouse에도 cross activity가 있나요?

    1 answer
    1. The cross-reactivity of this product in mice is not determined.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service