Skip to Content
Merck
All Photos(1)

Key Documents

E9750

Sigma-Aldrich

Estrone

≥99% (HPLC), powder, estrogen receptor agonist

Synonym(s):

1,3,5(10)-Estratrien-3-ol-17-one, 3-Hydroxy-1,3,5(10)-estratrien-17-one, Folliculin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22O2
CAS Number:
Molecular Weight:
270.37
Beilstein:
1915077
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Product Name

Estrone, ≥99%

biological source

synthetic (organic)

Quality Level

sterility

non-sterile

Assay

≥99%

form

powder

mp

258-260 °C (lit.)

solubility

dioxane: 50 mg/mL, clear, colorless to very faintly yellow

shipped in

ambient

storage temp.

room temp

SMILES string

C[C@]12CC[C@H]3[C@@H](CCc4cc(O)ccc34)[C@@H]1CCC2=O

InChI

1S/C18H22O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3,5,10,14-16,19H,2,4,6-9H2,1H3/t14-,15-,16+,18+/m1/s1

InChI key

DNXHEGUUPJUMQT-CBZIJGRNSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Estrone is a weak form of estrogen and exists as estrone sulphate. Estrone is a luteolytic estrogen produced by the corpus luteum. In the follicle, estrone is synthesized from androstenedione by the action of cytochrome P450 aromatase.

Application

Estrone has been used:
  • as medium supplement for hormone based degranulation studies of natural killer cells
  • as an endocrine disrupting compound for screening bacterial biosensor in toxic water.
  • as medium component for monitoring fatty acid synthase (FASN) activity in breast adenocarcinoma cell lines

Biochem/physiol Actions

Estrone is an agonist for the estrogen receptor. The estradiol to estrone interconversion is favourable in menopause. Oral hormone replacement therapy (HRT) of estradiol-17β increases circulating levels of estrone.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Nuclear Receptors (Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Lact. - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hormone replacement with estradiol: conventional oral doses result in excessive exposure to estrone
Friel PN, et al.
Alternative Medicine Review, 10(1), 36-42 (2005)
Estrone-a partial estradiol antagonist in the normal breast
Lundstrom E, et al.
Gynecological Endocrinology, 31(9), 747-749 (2015)
Pharmacology of estrogens and progestogens: influence of different routes of administration
Kuhl H
Climacteric : The Journal of the International Menopause Society, 8(1), 3-63 (2005)
The molecular control of corpus luteum formation, function, and regression
Stocco C, et al.
Endocrine Reviews, 28(1), 117-149 (2007)
Breast cancer metabolic cross-talk: Fibroblasts are hubs and breast cancer cells are gatherers of lipids
Lopes-Coelho F, et al.
Molecular and cellular endocrinology, 462(9), 93-106 (2018)

Articles

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service