Skip to Content
Merck
All Photos(1)

Key Documents

SAB4200119

Sigma-Aldrich

Monoclonal Anti-FLAG-Peroxidase antibody produced in rat

2-4 mg/mL, clone 6F7, purified immunoglobulin

Synonym(s):

Anti-ddddk, Anti-dykddddk

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.32

biological source

rat

conjugate

peroxidase conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6F7, monoclonal

form

buffered aqueous solution

species reactivity

all

concentration

2-4 mg/mL

technique(s)

western blot: 1:1,000-1:2,000 using extracts of transfected cells expressing C-terminal FLAG-tagged fusion protein

isotype

IgG1

immunogen sequence

DYKDDDDK

shipped in

dry ice

storage temp.

−20°C

General description

Monoclonal Anti-FLAG-Peroxidase is a purified immunoglobulin fraction of monoclonal Anti-FLAG (rat IgG1 isotype) isolated from culture supernatant of the 6F7 hybridoma cells grown in a bioreactor, conjugated to horseradish peroxidase (HRP). The hybridoma 6F7 was produced by the fusion of mouse myeloma cells and splenocytes from rat immunized with the FLAG peptide.
The product recognizes N-terminal, C-terminal, and internal FLAG-tagged fusion proteins. It is especially recommended for identifying C-terminal FLAG-tagged fusion proteins.

Immunogen

purified immunoglobulin fraction of monoclonal Anti-FLAG (rat IgG1 isotype) isolated from culture supernatant of the 6F7 hybridoma cells grown in a bioreactor, conjugated to horseradish peroxidase (HRP).

Application

Learn more product details in our FLAG® applications portal.
Monoclonal Anti-FLAG-Peroxidase, recognizes N-terminal, C-terminal and internal FLAG-tagged fusion proteins. The product is especially recommended for identifying C-terminal FLAG-tagged fusion proteins. The product can be used for immunoblotting.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 0.01% merthiolate.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jasjot Singh et al.
Nature communications, 13(1), 6212-6212 (2022-10-21)
Lysosomes are well-established as the main cellular organelles for the degradation of macromolecules and emerging as regulatory centers of metabolism. They are of crucial importance for cellular homeostasis, which is exemplified by a plethora of disorders related to alterations in
Jente Stouthamer et al.
Methods in molecular biology (Clifton, N.J.), 2690, 193-204 (2023-07-14)
Interactions between extracellular domains (ECDs) are crucial for many physiological processes in the cell, most importantly perception of its environment. However, studying these often-transient interactions can be challenging. Here we describe a method that allows for in vitro detection of
Claudia Isabelle Keller Valsecchi et al.
Nature, 589(7840), 137-142 (2020-11-20)
Confinement of the X chromosome to a territory for dosage compensation is a prime example of how subnuclear compartmentalization is used to regulate transcription at the megabase scale. In Drosophila melanogaster, two sex-specific non-coding RNAs (roX1 and roX2) are transcribed
DongHyuk Lee et al.
Nature communications, 11(1), 1838-1838 (2020-04-17)
Production of reactive oxygen species (ROS) is critical for successful activation of immune responses against pathogen infection. The plant NADPH oxidase RBOHD is a primary player in ROS production during innate immunity. However, how RBOHD is negatively regulated remains elusive.
Meidi Gu et al.
Nature immunology, 22(2), 193-204 (2021-01-06)
Metabolic reprograming toward aerobic glycolysis is a pivotal mechanism shaping immune responses. Here we show that deficiency in NF-κB-inducing kinase (NIK) impairs glycolysis induction, rendering CD8+ effector T cells hypofunctional in the tumor microenvironment. Conversely, ectopic expression of NIK promotes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service