Skip to Content
Merck
All Photos(3)

Key Documents

SAB4200071

Sigma-Aldrich

ANTI-FLAG® antibody, Rat monoclonal

clone 6F7, purified from hybridoma cell culture

Synonym(s):

Anti-ddddk, Anti-dykddddk

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.32

biological source

rat

conjugate

unconjugated

antibody form

purified from hybridoma cell culture
purified immunoglobulin

antibody product type

primary antibodies

clone

6F7, monoclonal

form

buffered aqueous solution

species reactivity

all

technique(s)

immunoprecipitation (IP): 2.5-5.0 μg using lysates of transiently transfected cells expressing C-terminal-FLAG-tagged protein
western blot: 0.5-1.0 μg/mL using extracts of transiently transfected cells expressing C-terminal-FLAG-tagged protein

isotype

IgG1

immunogen sequence

(DYKDDDDK)

shipped in

dry ice

storage temp.

−20°C

General description

Monoclonal Anti-FLAG® (rat IgG1 isotype) is derived from the hybridoma 6F7 produced by the fusion of mouse myeloma cells and splenocytes from rat immunized with the FLAG® peptide. The antibody is purified from culture supernatant of hybridoma cells grown in a bioreactor.

Monoclonal Anti-FLAG® recognizes N-terminal,
C-terminal and internal Flag-tagged fusion proteins. The product is especially recommended for identifying C-terminal FLAG®-tagged fusion proteins.
Epitope tags provide a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance, or poorly immunogenic proteins when protein specific antibodies are not available. Tagging with the FLAG® peptide sequence may be done at the N-terminus, N-terminus preceded by a methionine residue, C-terminus, or at internal positions of the target protein. FLAG may also be placed in associationith other tags. The small size of the FLAG® tag or sequence and its high hydrophilicity tend to decrease the possibility of interference with the protein expression, proteolytic maturation, antigenicity, and function.

The N-terminal FLAG® peptide sequence contains a unique enterokinase cleavage site allowing it to be completely removed from the purified fusion proteins. Cleavage of the C-terminal FLAG® peptide from a fusion protein catalyzed by Cu2+ ions has been reported. A sequence motif with five out of eight amino acid residues identical to the FLAG peptide is found in both rat and mouse Mg2+dependent protein b-phosphatase, as well as in the human and bovine enzyme.

Immunogen

FLAG peptide

DYKDDDDK

Application

ANTI-FLAG® antibody, Rat monoclonal has been used in:
  • chromatin immunoprecipitation (ChIP)
  • western blotting
  • coimmunoprecipitation
  • flow cytometric analysis

Browse additional application references in our FLAG® Literature portal.
Learn more product details in our FLAG® application portal.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Legal Information

ANTI-FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PHF21B overexpression promotes cancer stem cell-like traits in prostate cancer cells by activating the Wntbeta-catenin signaling pathway
Li Q, et al
Journal of Experimental & Clinical Cancer Research, 36(1), 85-85 (2017)
Xiaolei Li et al.
eLife, 6 (2017-08-19)
Acquired therapeutic resistance by tumors is a substantial impediment to reducing the morbidity and mortality that are attributable to human malignancies. The mechanisms responsible for the dramatic shift between chemosensitivity and chemoresistance in colorectal carcinoma have not been defined. Here
Rahul Bhowmick et al.
The Journal of biological chemistry, 287(42), 35004-35020 (2012-08-14)
Viruses have evolved to encode multifunctional proteins to control the intricate cellular signaling pathways by using very few viral proteins. Rotavirus is known to express six nonstructural and six structural proteins. Among them, NSP4 is the enterotoxin, known to disrupt
Upregulated expression of HOXB7 in intrahepatic cholangiocarcinoma is associated with tumor cell metastasis and poor prognosis
Dai L, et al.
Laboratory Investigation; a Journal of Technical Methods and Pathology, 99(6), 736-736 (2019)
Rotaviral enterotoxin nonstructural protein 4 targets mitochondria for activation of apoptosis during infection
Bhowmick R, et al.
The Journal of Biological Chemistry, 287(42), 35004-35020 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service