Skip to Content
Merck
All Photos(1)

Documents

S9396

Sigma-Aldrich

Sphingomyelinase from Bacillus cereus

buffered aqueous glycerol solution, ≥100 units/mg protein (Lowry)

Synonym(s):

Sphingomyelin choline phosphohydrolase, Sphingomyelin phosphodiesterase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

buffered aqueous glycerol solution

Quality Level

specific activity

≥100 units/mg protein (Lowry)

mol wt

34.2 kDa

foreign activity

Phospholipase C ≤1%

storage temp.

2-8°C

Gene Information

Bacillus cereus ATCC 10987 ... spH(2751530)

Looking for similar products? Visit Product Comparison Guide

General description

Sphingomyelinase (SMase) is a hydrolase, of 34.2 kDa and its active site contain as a minimum of two magnesium atoms.

Application

Sphingomyelinase from Bacillus cereus has been used:
  • in the study to analyse the effect of sphingomyelinase on Rubella virus infectivity in cell lines
  • in lipoprotein modification for the generation of cholesterol crystals in vitro
  • to aggregate LDL (low-density lipoprotein)

Sphingomyelinase has been used in a study to assess the interaction of actin with the HIV-1 accessory protein Nef. Sphingomyelinase has also been used in a study to investigate X-ray scattering as a quality-control tool for liposomal drug-delivery systems.

Biochem/physiol Actions

Initiates the formation of sphingomyelin-based second messengers. Activates MAPK (mitogen-activated protein kinase) and SAPKs (stress-activated protein kinases); generates ceramide from sphingomyelin.
Sphingomyelin phosphodiesterase (SMPD) participates in the metabolic reactions comprising sphingomyelin. SMPD has the ability to prevent colorectal carcinoma and colitis.

Unit Definition

One unit will hydrolyze 1.0 μmole of TNPAL-sphingomyelin per min at pH 7.4 at 37 °C.

Physical form

Solution in 50% glycerol containing 50 mM Tris-HCl, pH 7.5.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Pabst et al.
European biophysics journal : EBJ, 41(10), 915-929 (2012-06-05)
Biological membranes can be targets for compounds that either disrupt their barrier function or affect protein function via membrane-mediated processes. Biophysical studies on membrane-mimetic systems composed of membrane lipids have contributed substantially to our knowledge on the pertaining membrane physics
Analysis of the Effect of Sphingomyelinase on Rubella Virus Infectivity in Two Cell Lines
Otsuki N, et al.
Journal of virology (2017)
Antioxidants inhibit low density lipoprotein oxidation less at lysosomal pH: A possible explanation as to why the clinical trials of antioxidants might have failed
Ahmad F and Leake D S
Chemistry and Physics of Lipids, 213, 13-24 (2018)
Extracellular lipids accumulate in human carotid arteries as distinct three-dimensional structures and have proinflammatory properties
Lehti S, et al.
The American Journal of Pathology, 188(2), 525-538 (2018)
Raymond Tan et al.
Experimental and molecular pathology, 94(1), 51-57 (2012-06-23)
The HIV-1 accessory protein Nef is considered to play an important role in the development of a podocyte phenotype in HIV-1 associated nephropathy. We hypothesized that Nef may be altering the podocyte phenotype both structurally and functionally. To elucidate the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service