Skip to Content
Merck
All Photos(1)

Documents

S8633

Sigma-Aldrich

Sphingomyelinase from Staphylococcus aureus

buffered aqueous glycerol solution, 100-300 units/mg protein (Lowry)

Synonym(s):

Sphingomyelin choline phosphohydrolase, Sphingomyelin phosphodiesterase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32

form

buffered aqueous glycerol solution

Quality Level

specific activity

100-300 units/mg protein (Lowry)

storage temp.

2-8°C

Application

Sphingomyelinase from Staphylococcus aureus has been used to:
  • induce neurotoxicity in rat cortical cultures to study the protective effects of minocycline
  • determine the concentration of sphingomyelin from serum samples
  • enhance sphingomyelinase activity to study PARK9-mediated exosome biogenesis

Biochem/physiol Actions

Bacterial sphingomyelinase is active at neutral pH. When used in cell culture in vitro, it hydrolyzes the sphingomyelin on the outer leaflet of the plasma membrane and produces ceramide that is lipid-soluble. Sphingomyelinase is the key enzyme in the sphingomyelinase/ceramide pathway, which is implicated in the pathogenesis of several neurodegenerative disorders.
Initiates the formation of sphingomyelin-based second messengers. Activates MAPK (mitogen-activated protein kinase) and SAPKs (stress-activated protein kinases); generates ceramide from sphingomyelin.

Unit Definition

One unit will hydrolyze 1.0 μmol of TNPAL-sphingomyelin per min at pH 7.4 at 37 °C.

Physical form

Solution in 50% glycerol containing 0.25 M phosphate buffer, pH 7.5

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Measurement of sphingomyelin and ceramide cellular levels after sphingomyelinase-mediated sphingomyelin hydrolysis.
P Santana et al.
Methods in molecular biology (Clifton, N.J.), 105, 217-221 (1999-07-31)
Taiji Tsunemi et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(46), 15281-15287 (2014-11-14)
Kufor-Rakeb syndrome (KRS) is caused by loss-of-function mutations in ATP13A2 (PARK9) and characterized by juvenile-onset parkinsonism, pyramidal signs, and cognitive decline. Previous studies suggested that PARK9 deficiency causes lysosomal dysfunction and α-synuclein (α-syn) accumulation, whereas PARK9 overexpression suppresses toxicity of
Elin Rebecka Carlsson et al.
Frontiers in endocrinology, 9, 172-172 (2018-06-21)
Metabolic surgery is superior to lifestyle intervention in reducing weight and lowering glycemia and recently suggested as treatment for type 2 diabetes mellitus. Especially Roux-en-Y gastric bypass (RYGB) has been focus for much research, but still the mechanisms of action
Feixiang Wang et al.
The Journal of clinical investigation, 131(19) (2021-08-18)
Proper metabolic activities facilitate T cell expansion and antitumor function; however, the mechanisms underlying disruption of the T cell metabolic program and function in the tumor microenvironment (TME) remain elusive. Here, we show a zinc finger protein 91-governed (ZFP91-governed) mechanism
D I Kuz'menko et al.
Biomeditsinskaia khimiia, 58(5), 556-563 (2013-01-08)
The functional state of a sphingomyeline cycle and character of its mutual relations with the processes of free radical lipid oxidation during starvation of animals without any restriction of access to drinking water at 1, 2, 3 day (I phase)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service