Skip to Content
Merck
All Photos(1)

Documents

S5758

Sigma-Aldrich

Sodium tetrathionate dihydrate

≥98% (titration)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaO3SS2SO3Na · 2H2O
CAS Number:
Molecular Weight:
306.27
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.26

Assay

≥98% (titration)

solubility

H2O: 200 + 4mL H2O mg, clear to hazy, colorless to faintly yellow

density

2.1 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

O.O.[Na+].[Na+].[O-]S(=O)(=O)SSS([O-])(=O)=O

InChI

1S/2Na.H2O6S4.2H2O/c;;1-9(2,3)7-8-10(4,5)6;;/h;;(H,1,2,3)(H,4,5,6);2*1H2/q2*+1;;;/p-2

InChI key

HAEPBEMBOAIUPN-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • Gaussia princeps luciferase: a bioluminescent substrate for oxidative protein folding.: Sodium tetrathionate dihydrate is used to study its impact on the oxidative folding of proteins, utilizing Gaussia princeps luciferase as a bioluminescent substrate. This study advances understanding of protein chemistry and bioluminescence applications in biochemical assays (Yu T et al., 2018).

  • https://doi.org/10.2174/092986612800190973
    https://doi.org/10.1080/10826068.2011.544230
    https://doi.org/10.11150/kansenshogakuzasshi.83.380
  • A spectroscopic investigation into the reaction of sodium tetrathionate with cysteine.: The research utilizes spectroscopic methods to study the reaction between sodium tetrathionate dihydrate and cysteine, providing insights into the chemical interactions and potential applications in biochemistry and molecular biology (Church JS et al., 2008).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wriddhiman Ghosh et al.
Systematic and applied microbiology, 29(5), 396-403 (2006-07-11)
Paracoccus versutus-like isolates from the rhizosphere of Clitoria ternatea, a slender leguminous herb (family--Papilionaceae), found ubiquitously in waste places and village forests of the Lower Gangetic plains of India, presented a case of graduated infraspecific variation that was capped by
Gábor Peintler et al.
Physical chemistry chemical physics : PCCP, 12(10), 2356-2364 (2010-05-08)
It is experimentally proven that the stoichiometry of the tetrathionate-chlorite reaction is 2S4O(2-)6 + 8(1/2)ClO-(2) + 6H2O = 8SO(2-)4 + ClO-(3) + 7(1/2)Cl- + 12H+ near 1:4 molar ratio of the reactants. Re-evaluation of the previously measured front velocity--concentration curves
Mei Kikumoto et al.
Applied and environmental microbiology, 79(1), 113-120 (2012-10-16)
Thiosulfate dehydrogenase is known to play a significant role in thiosulfate oxidation in the acidophilic, obligately chemolithoautotroph, Acidithiobacillus ferrooxidans. Enzyme activity measured using ferricyanide as the electron acceptor was detected in cell extracts of A. ferrooxidans ATCC 23270 grown on
Poul Bennekou et al.
Blood cells, molecules & diseases, 48(2), 102-109 (2011-12-06)
The vanadate-induced increase in passive uptake of calcium and cobalt and their interference were studied in human red cells using (45)Ca and (57)Co as tracers. Vanadate is a potent inhibitor of the Ca-pump in red cells, although in fed cells
Bomba Dam et al.
Research in microbiology, 158(4), 330-338 (2007-05-19)
Chemolithotrophic oxidation of reduced sulfur compounds was studied in the betaproteobacterium Tetrathiobacter kashmirensis in correlation with its transposon (Tn5-mob)-inserted mutants impaired in sulfur oxidation (Sox(-)) and found to be carried out via the tetrathionate intermediate (S(4)I) pathway. The group of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service