Skip to Content
Merck
All Photos(1)

Key Documents

R9759

Sigma-Aldrich

Rat serum

Synonym(s):

Serum

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
biological source:
rat
sterility:
non-sterile
shipped in:
dry ice

biological source

rat

Quality Level

sterility

non-sterile

contains

preservative

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Rat serum is pooled from a normal donor population.

Application

Rat serum has been used as a blocking agent and controls in immunoassays.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Friederike Attig et al.
International journal of molecular sciences, 20(13) (2019-07-03)
(1) Background: Canine distemper virus (CDV)-induced demyelinating leukoencephalitis (CDV-DL) in dogs and Theiler's murine encephalomyelitis (TME) virus (TMEV)-induced demyelinating leukomyelitis (TMEV-DL) are virus-induced demyelinating conditions mimicking Multiple Sclerosis (MS). Reactive oxygen species (ROS) can induce the degradation of lipids and
J M Cook-Mills et al.
In vitro cellular & developmental biology. Animal, 32(3), 167-177 (1996-03-01)
Two long-term cultured cell lines were established from BALB/c mouse axillary and cervical lymph nodes that exhibited a combination of functional, morphological, and phenotypic characteristics consistent only with high endothelial venule cells. Spleen lymphocytes selectively bound and migrated across the
Charlotte L Scott et al.
Immunity, 49(2), 312-325 (2018-08-05)
Heterogeneity between different macrophage populations has become a defining feature of this lineage. However, the conserved factors defining macrophages remain largely unknown. The transcription factor ZEB2 is best described for its role in epithelial to mesenchymal transition; however, its role
Label-free protein detection using a microfluidic Coulter-counter device
Rodriguez-Trujillo R, et al.
Sensors and Actuators B, Chemical, 190, 922-927 (2014)
James Duffy et al.
Nanoscale, 10(26), 12797-12804 (2018-06-28)
Advances in prevention, diagnosis and therapy are coupled to innovation and development of new medical tools, leading to improved patient prognosis. We developed an automatic biosensor platform that could provide a non-invasive, rapid and personalised diagnosis using nanomechanical cantilever sensors.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service