Skip to Content
Merck
All Photos(4)

Key Documents

S4500

Sigma-Aldrich

L-Serine

≥99% (HPLC), ReagentPlus®

Synonym(s):

(S)-2-Amino-3-hydroxypropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
105.09
Beilstein:
1721404
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

Product Name

L-Serine, ReagentPlus®, ≥99% (HPLC)

product line

ReagentPlus®

Assay

≥99% (HPLC)

form

powder

color

white to off-white

mp

222 °C (dec.) (lit.)

solubility

H2O: 50 mg/mL, clear, colorless

application(s)

detection

storage temp.

room temp

SMILES string

N[C@@H](CO)C(O)=O

InChI

1S/C3H7NO3/c4-2(1-5)3(6)7/h2,5H,1,4H2,(H,6,7)/t2-/m0/s1

InChI key

MTCFGRXMJLQNBG-REOHCLBHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-Serine has been used in the preparation of Tris-BSAN buffer for homogenization. It has also been used for quantitative analysis of excretion of polypeptides in normal urine.[1][2]

Biochem/physiol Actions

L-Serine is a non-essential amino acid that acts as a precursor for nucleotide synthesis. It has roles in the development and function of the central nervous system.[3] L-Serine also has a role in cellular proliferation.[4]
Precursor of glycine by serine hydroxymethyltransferase.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Taiga Okumura et al.
Scientific reports, 9(1), 3520-3520 (2019-03-07)
Radiocaesium-bearing microparticles (CsMPs) composed of silicate glass were released by the Fukushima Daiichi Nuclear Power Plant accident in March 2011. Since CsMPs contain a high concentration of radiocaesium, their dynamics and fate in the environment are urgent issues to be
Analysis of GSH and GSSG after derivatization with N-ethylmaleimide
Nature Protocols, 8(9) (2003)
Abel Lajtha, Simo S. Oja, Pirjo Saransaari, Arne Schousboe
Handbook of Neurochemistry and Molecular Neurobiology: Amino Acids and Peptides in the Nervous System , 4 (2007)
Quantitative amino acid and proteomic analysis: Very low excretion of polypeptides >750 Da in normal urine
Anthony G.W.Norden
Kidney International, 66(5) (2004)
Taiga Okumura et al.
Scientific reports, 8(1), 9707-9707 (2018-06-28)
Radiocesium-bearing microparticles (CsPs) substantially made of silicate glass are a novel form of radiocesium emitted from the broken containment vessel of Fukushima Dai-ichi nuclear power plant. CsPs have a potential risk of internal radiation exposure caused by inhalation. Radiation-contaminated waste

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service