Skip to Content
Merck
All Photos(5)

Documents

G1251

Sigma-Aldrich

L-Glutamic acid

≥99% (HPLC), suitable for microbiological culture, ReagentPlus®

Synonym(s):

(S)-2-Aminopentanedioic acid, Glu

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
147.13
Beilstein:
1723801
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

product name

L-Glutamic acid, ReagentPlus®, ≥99% (HPLC)

Agency

suitable for SM 5210

product line

ReagentPlus®

Assay

≥99% (HPLC)

form

powder

technique(s)

microbiological culture: suitable

color

white to off-white

mp

205 °C (dec.) (lit.)

solubility

1 M HCl: soluble 100 mg/mL

density

1.54 g/cm3 at 20 °C

cation traces

C: 40.4-41.2%
N: 9.2-9.8%

SMILES string

N[C@@H](CCC(O)=O)C(O)=O

InChI

1S/C5H9NO4/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H,7,8)(H,9,10)/t3-/m0/s1

InChI key

WHUUTDBJXJRKMK-VKHMYHEASA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-Glutamic acid has been used to enhance endogenous acetylcholine overflow. It has been used as a supplement in the growth medium of meat-Spoiling Pseudomonas fragi 72.

Biochem/physiol Actions

An excitatory amino acid neurotransmitter that is an agonist at all subtypes of glutamate receptors (metabotropic, kainate, NMDA, and AMPA).
Glutamic acid, or glutamate, the salt form of glutamic acid, functions as a neurotransmitter and also serves as a precursor to other neurotransmitters such as γ-aminobutyric acid. Glutamic acid also plays a key role in many metabolic pathways, that controls growth, reproduction, maintenance and immunity. It is converted to α-ketoglutarate, a key component of the TCA (tricarboxylic acid) cycle, and a precursor for the biosynthesis of nucleic acids and certain amino acids. In cells, glutamine is converted to glutamate by the enzyme glutaminase.
Glutamine serves as a source of energy for rapidly dividing cells comprising lymphocytes, enterocytes, macrophages and tumors. Glutamine mediates protein turnover via cellular mTOR (mammalian target of rapamycin) signaling. It is also known to be associated with the inhibition of apoptosis.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glutamic acid, twenty years later.
Garattini S
The Journal of Nutrition, 130, 901S-909S (2000)
Mixed carbon source utilization of meat-spoiling Pseudomonas fragi 72 in relation to oxygen limitation and carbon dioxide inhibition.
Molin G.
Applied and Environmental Microbiology, 49(6), 1442-1447 (1985)
The biology of cancer: metabolic reprogramming fuels cell growth and proliferation.
DeBerardinis RJ
Cell Metabolism, 7(1), 11-20 (2008)
Cristina Giaroni et al.
European journal of pharmacology, 476(1-2), 63-69 (2003-09-13)
Several reports suggest that enteric cholinergic neurons are subject to a tonic inhibitory modulation, whereas few studies are available concerning the role of facilitatory pathways. Glutamate, the main excitatory neurotransmitter in the central nervous system (CNS), has recently been described
Jasdeep Saini et al.
Journal of clinical medicine, 9(10) (2020-10-15)
In many neurodegenerative and muscular disorders, and loss of innervation in sarcopenia, improper reinnervation of muscle and dysfunction of the motor unit (MU) are key pathogenic features. In vivo studies of MUs are constrained due to difficulties isolating and extracting

Articles

Amino Acid Codon Wheel for fast RNA translation. Find which amino acid is translated from your RNA sequence quickly and easily.

Amino Acid Codon Wheel for fast RNA translation. Find which amino acid is translated from your RNA sequence quickly and easily.

Amino Acid Codon Wheel for fast RNA translation. Find which amino acid is translated from your RNA sequence quickly and easily.

Amino Acid Codon Wheel for fast RNA translation. Find which amino acid is translated from your RNA sequence quickly and easily.

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Chromatograms

application for HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service