Skip to Content
Merck
All Photos(2)

Key Documents

S2404

Sigma-Aldrich

Sodium acetate buffer solution

pH 7.0±0.05 (25 °C), BioXtra, for molecular biology, 3 M, non-sterile; 0.2 μm filtered

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
3786729
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.52

grade

for molecular biology

Quality Level

sterility

non-sterile; 0.2 μm filtered

product line

BioXtra

form

liquid

concentration

3 M

pH

7.0±0.05 (25 °C)

suitability

suitable for DNA precipitation

storage temp.

2-8°C

SMILES string

[Na+].CC(O)=O.CC([O-])=O

InChI

1S/2C2H4O2.Na/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+1/p-1

InChI key

BHZOKUMUHVTPBX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sigma′s Sodium Acetate Buffer is a 3M solution prepared with 18 megaohm water and filter-sterilized.

Application

Sodium acetate buffer solution has been used for the precipitation of DNA. It has also been used as a component of supporting electrolyte for electrochemical sensor.
Sodium acetate buffer solution is suitable for elution of proteins and for use in general molecular biology applications.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Buket Onel et al.
Methods in molecular biology (Clifton, N.J.), 2035, 201-222 (2019-08-25)
DNA G-quadruplexes are globular nucleic acid secondary structures which occur throughout the human genome under physiological conditions. There is accumulating evidence supporting G-quadruplex involvement in a number of important aspects of genome functions, including transcription, replication, and genomic stability, and
Application of sensitive electrochemical sensing system for detecting bromate from disinfection process in desalination plant
Lee YG and Jang A
Desalination, 423, 135-140 (2017)
Sambrook, J., and Russell, D.W.
Molecular Cloning: A Laboratory Manual, A1-A1 (2001)
Chun-Chun Chen et al.
Journal of visualized experiments : JoVE, (62)(62), doi:10-doi:10 (2012-05-09)
Knowing the timing, level, cellular localization, and cell type that a gene is expressed in contributes to our understanding of the function of the gene. Each of these features can be accomplished with in situ hybridization to mRNAs within cells.
Jaspreet Kaur et al.
Antimicrobial agents and chemotherapy, 55(2), 659-666 (2010-12-01)
In this study we utilized the concept of rational drug design to identify novel compounds with optimal selectivity, efficacy and safety, which would bind to the target enzyme pteridine reductase 1 (PTR1) in Leishmania parasites. Twelve compounds afforded from Baylis-Hillman

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service