P2069 is P3803 supplied with an equilibration buffer. Equilibration buffer B5658-1EA is required for DNA extraction.
P3803
Phenol:Chloroform:Isoamyl Alcohol 25:24:1, Saturated with 10mM Tris, pH 8.0, 1mM EDTA
for molecular biology
Synonym(s):
DNA extraction
About This Item
Recommended Products
grade
for molecular biology
Quality Level
Assay
≥99.0%
form
liquid
pH
6.5-6.9(pH of phenolic phase)
application(s)
agriculture
storage temp.
2-8°C
Application
Warning
Other Notes
Recommended products
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Muta. 2 - Repr. 2 - Skin Corr. 1B - STOT RE 1 Oral - STOT RE 2 - STOT SE 3
Target Organs
Central nervous system, Liver,Kidney, Nervous system,Kidney,Liver,Skin
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
Hello, I have used in the past P2069, which needs the equilibration buffer for pH8. Does P3803 also need the same equilibration buffer B5658-1EA? Thank you!
1 answer-
Helpful?
-
-
I received two bottles. One for the phenol and one for the buffer. Do I need to add the phenol to the buffer? or the buffer already contains the phenol?
1 answer-
Yes, the equilibration buffer is to be added before use, according to the following instructions:
Add the entire contents of the small bottle of Equilibration Buffer to the large bottle of Phenol:Chloroform:Isoamyl Alcohol reagent.
Mix gently and allow the phases to separate before use, typically 2–4 hours.
This will adjust the pH of the phenol phase from pH 6.7±0.2 to 8.0±0.2.Please see the link below to review the product datasheet for additional information:
https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/490/315/p3803pis.pdfHelpful?
-
-
What is the Department of Transportation shipping information for this product?
1 answer-
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Helpful?
-
-
I see that you list two equilibrated Phenol:Chloroform:Isoamyl alcohol products. When should I use Product P3803, and when should I use Product P2069?
1 answer-
In short, P3803 is for RNA work, and P2069 is for DNA work. Product P2069 consists of one bottle of Product P3803 and one bottle of Product B5658. The B5658 is added to the P3803 to raise the pH of the organic phase from 6.7 to 8.0; pH 8.0 is not suitable for RNA work, because RNA can degrade at the higher pH.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service