Skip to Content
Merck
All Photos(1)

Key Documents

71196

Sigma-Aldrich

Sodium acetate solution

BioUltra, for molecular biology, ~3 M in H2O

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H3NaO2
CAS Number:
Molecular Weight:
82.03
Beilstein:
3595639
MDL number:
UNSPSC Code:
12161700
eCl@ss:
39021906
PubChem Substance ID:
NACRES:
NA.21

grade

Molecular Biology
for molecular biology

Quality Level

product line

BioUltra

form

liquid

concentration

~3 M in H2O

technique(s)

precipitation assay: suitable

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected

pH

7.0-8.0 (25 °C, 3 M in H2O)

anion traces

chloride (Cl-): ≤10 mg/kg
phosphate (PO43-): ≤5 mg/kg
sulfate (SO42-): ≤20 mg/kg

cation traces

Al: ≤1 mg/kg
As: ≤0.1 mg/kg
Ba: ≤1 mg/kg
Bi: ≤1 mg/kg
Ca: ≤5 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤50 mg/kg
Li: ≤1 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Mo: ≤1 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Sr: ≤1 mg/kg
Zn: ≤1 mg/kg

λ

3 M in H2O

UV absorption

λ: 260 nm Amax: ≤0.030
λ: 280 nm Amax: ≤0.024

SMILES string

[Na+].CC([O-])=O

InChI

1S/C2H4O2.Na/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

VMHLLURERBWHNL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium acetate solution has been used:
  • in DNA extraction
  • in reverse crosslinking and DNA purification
  • to precipitate Gata6 (GATA binding protein 6) cDNA as a part of high-purity plasmid preparation

Biochem/physiol Actions

Sodium acetate serves as a fixed base. It is useful to rectify acidosis among acidotic patients. Orginally, sodium acetate is used as dialysate for hemodialysis, but is known to cause cardiovascular instability.

Other Notes

Basic method of DNA precipitation with ethanolcitation
Basic method of DNA precipitation with ethanol

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sodium acetate infusion in critically ill trauma patients for hyperchloremic acidosis
McCague A, et al.
Scandinavian Journal of Trauma, Resuscitation and Emergency Medicine , 19(1), 24-24 (2011)
Genome-wide mapping of DNase I hypersensitive sites in plants
Plant Functional Genomics, 71-89 (2015)
An ultra-low-input native ChIP-seq protocol for genome-wide profiling of rare cell populations
Brind Amour J, et al.
Nature Communications, 6, 6033-6033 (2015)
Derivation of extraembryonic endoderm stem (XEN) cells from mouse embryos and embryonic stem cells
Niakan KK, et al.
Nature Protocols, 8(6), 1028-1028 (2013)
D.M. Wallace
Methods in Enzymology, 152, 42-42 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service