Skip to Content
Merck
All Photos(1)

Key Documents

Q2763

Sigma-Aldrich

Quinalizarin

≥95% (HPLC), powder, plasmodium falciparum casein kinase 2α (PfCK2α) inhibitor

Synonym(s):

1,2,5,8-Tetrahydroxy-9,10-anthraquinone, 1,2,5,8-Tetrahydroxyanthraquinone, Alizarin Bordeaux BD, Alizarinbordeaux, Alizarine Bordeaux, Alizarine Bordeaux B, C.I. 58500, C.I. Mordant Violet 26, Khinalizarin, NSC 144046, NSC 4896, PHF 016

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H8O6
CAS Number:
Molecular Weight:
272.21
Colour Index Number:
58500
Beilstein:
1889617
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.77

Product Name

Quinalizarin, ≥95% (HPLC)

Assay

≥95% (HPLC)

form

powder

color

red

mp

≥300 °C

storage temp.

room temp

SMILES string

Oc1ccc2C(=O)c3c(O)ccc(O)c3C(=O)c2c1O

InChI

1S/C14H8O6/c15-6-3-4-7(16)11-10(6)12(18)5-1-2-8(17)13(19)9(5)14(11)20/h1-4,15-17,19H

InChI key

VBHKTXLEJZIDJF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Quinalizarin has been used:
  • as a casein kinase II (CKII) inhibitor to study its effect on CKII-mediated phosphorylation of importin α on subcellular scaling in sperm chromosomes and egg extract
  • as a CKII inhibitor to study its ability to block parasite multiplication in a [3H]-hypoxanthine incorporation assay
  • to study its effect on colorectal cancer (CRC) cell cycle arrest, cell apoptosis, and reactive oxygen species (ROS) generation in SW480 and HCT-116 cell lines

Biochem/physiol Actions

Quinalizarin is a potent (IC50 = 110 nM), ATP-competitive, and highly selective (IC50 >1μM against CK1 and 72 other kinases) casein Kinase II (CK2) inhibitor.
Quinalizarin is a potent, ATP-competitive, and highly selective casein Kinase II (CK2) inhibitor.
Studies show that quinalizarin can prevent the in vitro activity of recombinant Plasmodium falciparum casein kinase 2α (PfCK2α) catalytic subunit with an IC50 of 2μM.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammad S Zaman et al.
Metallomics : integrated biometal science, 8(1), 82-90 (2015-12-02)
Protein kinase CK2 is a pleiotropic tetrameric enzyme, regulating numerous biological processes from cell proliferation to stress response. This study demonstrates for the first time that CK2 is involved in the regulation of metal uptake and toxicity in neuronal cells.
Qianwen Li et al.
Biological research, 52(1), 22-22 (2019-04-18)
Tumor microenvironment (TME) plays a vital role in determining the outcomes of radiotherapy. As an important component of TME, vascular endothelial cells are involved in the perivascular resistance niche (PVRN), which is formed by inflammation or cytokine production induced by
Ling-Qi Meng et al.
Medical science monitor : international medical journal of experimental and clinical research, 24, 3710-3719 (2018-06-04)
BACKGROUND Quinalizarin (1,2,5,8-tetrahydroxyanthraquinone) exhibits potentially useful anticancer effects by inducing apoptosis in several types of cancer, but its underlying mechanism of action remains unknown. The present study examined the effects of quinalizarin on the induction of cell cycle arrest, apoptosis
Y Zhou et al.
Indian journal of cancer, 52 Suppl 2, e119-e124 (2016-01-06)
Protein kinase CK2 is widely expressed in eukaryotic cells, and plays an important role in cell proliferation, migration, apoptosis, etc. The aim of the current study is to explore how Quinalizarin, a specific CK2 inhibitor, affects the cell proliferation, migration
Yinpeng Jin et al.
EBioMedicine, 34, 231-242 (2018-08-06)
It has previously been reported that human adipose-derived stem cells (hASCs) can promote the regeneration of damaged tissues in rats with liver failure through a 'paracrine effect'. Here we demonstrate a therapeutic effect of hASCs derived Extracellular Vesicles (EVs) on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service